Assay Details
| Gene Symbol: EDAR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022336.3 | NP_071731.1 | 1 - 2 | 412 | 60 | |
| XM_006712204.1 | XP_006712267.1 | 1 - 2 | 296 | 60 | ||
| GenBank mRNA | AF130988.1 | - | 1 - 2 | 413 | 60 | |
| AK296936.1 | - | 1 - 2 | 206 | 60 | ||
| AK313781.1 | - | 1 - 2 | 296 | 60 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02002183.hg
Additional Information:
This assay is in the 5'UTR region of the AF130988.1,AK296936.1,AK313781.1,NM_022336.3,XM_006712204.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
hair follicle development
apoptotic process
epidermis development
positive regulation of gene expression
cell differentiation
tumor necrosis factor-mediated signaling pathway
positive regulation of NF-kappaB import into nucleus
odontogenesis of dentin-containing tooth
positive regulation of I-kappaB kinase/NF-kappaB signaling
pigmentation
positive regulation of JNK cascade
salivary gland cavitation
receptor activity
transmembrane signaling receptor activity
protein binding
hair follicle development
apoptotic process
epidermis development
positive regulation of gene expression
cell differentiation
tumor necrosis factor-mediated signaling pathway
positive regulation of NF-kappaB import into nucleus
odontogenesis of dentin-containing tooth
positive regulation of I-kappaB kinase/NF-kappaB signaling
pigmentation
positive regulation of JNK cascade
salivary gland cavitation
receptor activity
transmembrane signaling receptor activity
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02035464.hg.1 | TC02002183.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02035464.hg.1 | ENST00000258443.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02035464.hg.1 | ENST00000376651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02035464.hg.1 | ENST00000409271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02035464.hg.1 | NM_022336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02035468.hg.1 | TC02002183.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02035468.hg.1 | ENST00000258443.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02035468.hg.1 | ENST00000376651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02035468.hg.1 | NM_022336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02035463.hg.1 | TC02002183.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02035463.hg.1 | ENST00000258443.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02035463.hg.1 | ENST00000376651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02035463.hg.1 | ENST00000409271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02035463.hg.1 | NM_022336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02035467.hg.1 | TC02002183.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02035467.hg.1 | ENST00000258443.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02035467.hg.1 | ENST00000376651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02035467.hg.1 | ENST00000409271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02035467.hg.1 | NM_022336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | ENST00000258443.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | ENST00000258443.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | ENST00000258443.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | ENST00000258443.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | ENST00000258443.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | ENST00000258443.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | ENST00000258443.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | ENST00000258443.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | ENST00000258443.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | ENST00000258443.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | ENST00000258443.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | ENST00000258443.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | ENST00000258443.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | ENST00000258443.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | ENST00000376651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | ENST00000376651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | ENST00000376651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | ENST00000376651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | ENST00000376651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | ENST00000376651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | ENST00000376651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | ENST00000376651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | ENST00000376651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | ENST00000376651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | ENST00000376651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | ENST00000376651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | ENST00000376651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | ENST00000376651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | ENST00000376651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | ENST00000409271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | ENST00000409271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | ENST00000409271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | ENST00000409271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | ENST00000409271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | ENST00000409271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | ENST00000409271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | ENST00000409271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | ENST00000409271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | ENST00000409271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | ENST00000409271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | ENST00000409271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | ENST00000409271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | ENST00000409271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | ENST00000409271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | NM_022336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | NM_022336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | NM_022336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | NM_022336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | NM_022336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | NM_022336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | NM_022336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | NM_022336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | NM_022336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | NM_022336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | NM_022336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | NM_022336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | NM_022336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013815.hg.1 | NM_022336.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200201176.hg.1 | ENST00000258443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200201176.hg.1 | ENST00000376651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200201176.hg.1 | ENST00000409271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200201176.hg.1 | NM_022336.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200201176.hg.1 | ENST00000258443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200201176.hg.1 | ENST00000376651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200201176.hg.1 | ENST00000409271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200201176.hg.1 | NM_022336.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200201176.hg.1 | TC0200013815.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200201177.hg.1 | ENST00000258443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200201177.hg.1 | ENST00000376651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200201177.hg.1 | NM_022336.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200201177.hg.1 | ENST00000258443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200201177.hg.1 | ENST00000376651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200201177.hg.1 | NM_022336.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200201177.hg.1 | TC0200013815.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200201171.hg.1 | ENST00000258443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200201171.hg.1 | ENST00000376651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200201171.hg.1 | ENST00000409271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200201171.hg.1 | NM_022336.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200201171.hg.1 | ENST00000258443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200201171.hg.1 | ENST00000376651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200201171.hg.1 | ENST00000409271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200201171.hg.1 | NM_022336.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200201171.hg.1 | TC0200013815.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200201172.hg.1 | ENST00000258443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200201172.hg.1 | ENST00000376651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200201172.hg.1 | ENST00000409271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200201172.hg.1 | NM_022336.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200201172.hg.1 | ENST00000258443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200201172.hg.1 | ENST00000376651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200201172.hg.1 | ENST00000409271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200201172.hg.1 | NM_022336.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200201172.hg.1 | TC0200013815.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | ENST00000258443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | ENST00000258443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | ENST00000258443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | ENST00000258443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | ENST00000258443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | ENST00000258443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | ENST00000258443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | ENST00000258443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | ENST00000258443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | ENST00000258443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | ENST00000258443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | ENST00000258443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | ENST00000258443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | ENST00000258443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | ENST00000376651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | ENST00000376651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | ENST00000376651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | ENST00000376651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | ENST00000376651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | ENST00000376651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | ENST00000376651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | ENST00000376651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | ENST00000376651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | ENST00000376651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | ENST00000376651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | ENST00000376651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | ENST00000376651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | ENST00000376651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | ENST00000376651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | ENST00000409271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | ENST00000409271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | ENST00000409271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | ENST00000409271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | ENST00000409271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | ENST00000409271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | ENST00000409271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | ENST00000409271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | ENST00000409271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | ENST00000409271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | ENST00000409271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | ENST00000409271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | ENST00000409271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | ENST00000409271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | ENST00000409271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | NM_022336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | NM_022336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | NM_022336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | NM_022336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | NM_022336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | NM_022336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | NM_022336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | NM_022336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | NM_022336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | NM_022336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | NM_022336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | NM_022336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | NM_022336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013815.hg.1 | NM_022336.3 | Refseq |

EDAR
Genomic Map