Assay Details
| Gene Symbol: CMTM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AL832450.1 | - | 1 | 1728 | 63 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AA601968.1 | - | 1 | Chr. 3 - 32393363 - 32393627 [-] on Build GRCh38 | 63 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | CKLF like MARVEL transmembrane domain containing 8 | CMTM8 |
CKLFSF8, CKLFSF8-V2 |
-21.4kb | Sense | Chr. 3 - 32238679 - 32370325 on NCBI Build GRCh38 |
| Downstream | CKLF like MARVEL transmembrane domain containing 6 | CMTM6 |
CKLFSF6, PRO2219 |
87.5kb | Anti-sense | Chr. 3 - 32481312 - 32502911 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

CMTM7
Genomic Map