Assay Details
| Gene Symbol: FGD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004463.2 | NP_004454.2 | 9 - 10 | 2433 | 78 | |
| GenBank mRNA | AK226117.1 | - | 9 - 10 | 2223 | 78 | |
| AK299635.1 | - | 7 - 8 | 1142 | 78 | ||
| BC034530.1 | - | 9 - 10 | 1945 | 4938457 | 78 | |
| U11690.1 | - | 9 - 10 | 2429 | 78 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X001074.hg.1
Set Membership: |
|

FGD1
Genomic Map