Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
ENST00000437377.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
ENST00000437377.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
ENST00000606775.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
ENST00000606775.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
ENST00000606775.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
ENST00000606775.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
ENST00000606775.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
ENST00000606775.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
ENST00000606775.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
ENST00000606775.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
ENST00000606775.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
ENST00000606775.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
ENST00000606775.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
ENST00000606775.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
ENST00000606775.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
ENST00000606775.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
ENST00000606775.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
ENST00000437377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
ENST00000437377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
ENST00000606775.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
ENST00000606775.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
ENST00000606775.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
ENST00000606775.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
ENST00000606775.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
ENST00000606775.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
ENST00000606775.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
ENST00000606775.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
ENST00000606775.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
ENST00000606775.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
ENST00000606775.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
ENST00000606775.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
ENST00000606775.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
ENST00000606775.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
ENST00000606775.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015585.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100216416.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100216416.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100216416.hg.1 |
ENST00000619932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100216416.hg.1 |
NR_104217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100216416.hg.1 |
TC0100015585.hg.1 |
Clariom D TC |