Assay Details
| Gene Symbol: ABCB11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003742.2 | NP_003733.2 | 1 - 2 | 94 | 129 | |
| XM_006712817.3 | XP_006712880.1 | 1 - 2 | 164 | 129 | ||
| XM_011512077.2 | XP_011510379.1 | 1 - 2 | 164 | 129 | ||
| XM_011512078.2 | XP_011510380.1 | 1 - 2 | 164 | 129 | ||
| XM_011512080.2 | XP_011510382.1 | 1 - 2 | 167 | 129 | ||
| XM_017005165.1 | XP_016860654.1 | 1 - 2 | 166 | 129 | ||
| GenBank mRNA | AF091582.1 | - | 1 - 2 | 94 | 129 | |
| AF136523.1 | - | 1 - 2 | 95 | 129 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02002508.hg
Additional Information:
This assay is in the 5'UTR region of the AF091582.1,AF136523.1,NM_003742.2,XM_006712817.3,XM_011512077.2,XM_011512078.2,XM_011512080.2,XM_017005165.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
bile acid biosynthetic process
transport
bile acid and bile salt transport
canalicular bile acid transport
sodium ion transmembrane transport
transporter activity
ATP binding
sodium-exporting ATPase activity, phosphorylative mechanism
canalicular bile acid transmembrane transporter activity
bile acid-exporting ATPase activity
bile acid biosynthetic process
transport
bile acid and bile salt transport
canalicular bile acid transport
sodium ion transmembrane transport
transporter activity
ATP binding
sodium-exporting ATPase activity, phosphorylative mechanism
canalicular bile acid transmembrane transporter activity
bile acid-exporting ATPase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02039723.hg.1 | TC02002508.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02039723.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02039723.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02039724.hg.1 | TC02002508.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02039724.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02039724.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000478354.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000478354.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000478354.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000478354.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200208357.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200208357.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200208357.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200208357.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200208357.hg.1 | TC0200014848.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200208359.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200208359.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200208359.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200208359.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200208359.hg.1 | TC0200014848.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000478354.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000478354.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000478354.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000478354.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200208358.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200208358.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200208358.hg.1 | ENST00000263817.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200208358.hg.1 | NM_003742.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200208358.hg.1 | TC0200014848.hg.1 | Clariom D TC |

ABCB11
Genomic Map