Assay Details
| Gene Symbol: ABCB11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003742.2 | NP_003733.2 | 2 - 3 | 191 | 143 | |
| GenBank mRNA | AF091582.1 | - | 2 - 3 | 191 | 143 | |
| AF136523.1 | - | 2 - 3 | 192 | 143 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF091582.1, AF136523.1, NM_003742.2 transcripts.
Set Membership: |
|

ABCB11
Genomic Map