Assay Details
| Gene Symbol: DCAF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286810.1 | NP_001273739.1 | 3 - 4 | 409 | 147 | |
| NM_024345.4 | NP_077321.3 | 3 - 4 | 935 | 147 | ||
| XM_005251577.3 | XP_005251634.1 | 3 - 4 | 956 | 147 | ||
| GenBank mRNA | AK074532.1 | - | 3 - 4 | 422 | 147 | |
| AK309712.1 | - | 3 - 4 | 409 | 147 | ||
| AK315756.1 | - | 3 - 4 | 388 | 147 | ||
| BC011959.1 | - | 1 - 2 | 138 | 147 | ||
| BC108686.1 | - | 3 - 4 | 904 | 6157119 | 147 | |
| BC139834.1 | - | 3 - 4 | 422 | 147 | ||
| CD299973.1 | - | 3 - 4 | 450 | 147 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000218.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09002594.hg.1 | TC09000218.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002594.hg.1 | ENST00000242323.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002594.hg.1 | ENST00000377724.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002594.hg.1 | ENST00000483167.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002594.hg.1 | NM_024345.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000452964.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000478453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000478453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000478453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000478453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002593.hg.1 | TC09000218.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002593.hg.1 | ENST00000242323.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002593.hg.1 | ENST00000377724.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002593.hg.1 | NM_024345.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900148914.hg.1 | ENST00000242323.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148914.hg.1 | ENST00000377724.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148914.hg.1 | ENST00000461549.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148914.hg.1 | ENST00000483167.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148914.hg.1 | NM_001286810.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900148914.hg.1 | NM_024345.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900148914.hg.1 | ENST00000242323.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148914.hg.1 | ENST00000377724.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148914.hg.1 | ENST00000461549.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148914.hg.1 | ENST00000483167.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148914.hg.1 | NM_001286810.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900148914.hg.1 | NM_024345.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900148914.hg.1 | TC0900007167.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000452964.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000478453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000478453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000478453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000478453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900148913.hg.1 | ENST00000242323.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148913.hg.1 | ENST00000377724.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148913.hg.1 | ENST00000461549.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148913.hg.1 | NM_001286810.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900148913.hg.1 | NM_024345.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900148913.hg.1 | ENST00000242323.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148913.hg.1 | ENST00000377724.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148913.hg.1 | ENST00000461549.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148913.hg.1 | NM_001286810.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900148913.hg.1 | NM_024345.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900148913.hg.1 | TC0900007167.hg.1 | Clariom D TC |

DCAF10
Genomic Map