| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009846.hg.1 |
ENST00000379731.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009846.hg.1 |
ENST00000379731.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009846.hg.1 |
ENST00000379731.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009846.hg.1 |
ENST00000379731.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009846.hg.1 |
ENST00000379731.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009846.hg.1 |
ENST00000379731.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009846.hg.1 |
ENST00000379731.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009846.hg.1 |
ENST00000379731.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009846.hg.1 |
ENST00000535206.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009846.hg.1 |
ENST00000535206.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009846.hg.1 |
ENST00000535206.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009846.hg.1 |
ENST00000535206.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009846.hg.1 |
NM_001497.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009846.hg.1 |
NM_001497.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009846.hg.1 |
NM_001497.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009846.hg.1 |
NM_001497.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009846.hg.1 |
NM_001497.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009846.hg.1 |
NM_001497.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009846.hg.1 |
NM_001497.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009846.hg.1 |
NM_001497.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09013216.hg.1 |
TC09001008.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09013216.hg.1 |
ENST00000379731.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09013216.hg.1 |
ENST00000535206.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09013216.hg.1 |
NM_001497.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
ENST00000426270.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
ENST00000426270.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
ENST00000426270.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
ENST00000426270.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
ENST00000426270.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
ENST00000426270.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
ENST00000426270.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
ENST00000426270.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
ENST00000426270.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
ENST00000426270.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
ENST00000426270.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
ENST00000426270.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
ENST00000442432.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
ENST00000442432.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
ENST00000442432.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
ENST00000442432.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
ENST00000442432.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
ENST00000442432.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
ENST00000442432.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
ENST00000442432.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
ENST00000442432.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
NR_108108.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
NR_108108.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
NR_108108.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
NR_108108.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
NR_108108.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
NR_108108.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
NR_108108.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
NR_108108.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
NR_108108.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
NR_108108.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
NR_108108.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
NR_108109.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
NR_108109.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
NR_108109.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
NR_108109.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
NR_108109.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
NR_108109.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
NR_108109.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
NR_108109.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
NR_108109.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
NR_108109.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
NR_108109.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
NR_108110.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
NR_108110.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
NR_108110.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
NR_108110.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
NR_108110.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
NR_108110.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
NR_108110.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006966.hg.1 |
NR_108110.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09013214.hg.1 |
TC09001008.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09013214.hg.1 |
ENST00000379731.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09013214.hg.1 |
ENST00000535206.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09013214.hg.1 |
ENST00000541701.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09013214.hg.1 |
NM_001497.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09013215.hg.1 |
TC09001008.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09013215.hg.1 |
ENST00000379731.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09013215.hg.1 |
ENST00000535206.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09013215.hg.1 |
ENST00000541701.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09013215.hg.1 |
NM_001497.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09013217.hg.1 |
TC09001008.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09013217.hg.1 |
ENST00000379731.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09013217.hg.1 |
ENST00000535206.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09013217.hg.1 |
ENST00000541701.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09013217.hg.1 |
NM_001497.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09013218.hg.1 |
TC09001008.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09013218.hg.1 |
ENST00000379731.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09013218.hg.1 |
ENST00000535206.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09013218.hg.1 |
NM_001497.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900167369.hg.1 |
ENST00000379731.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900167369.hg.1 |
ENST00000535206.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900167369.hg.1 |
NM_001497.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900167369.hg.1 |
ENST00000379731.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900167369.hg.1 |
ENST00000535206.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900167369.hg.1 |
NM_001497.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900167369.hg.1 |
TC0900009846.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0900009846.hg.1 |
ENST00000379731.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009846.hg.1 |
ENST00000379731.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009846.hg.1 |
ENST00000379731.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009846.hg.1 |
ENST00000379731.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009846.hg.1 |
ENST00000379731.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009846.hg.1 |
ENST00000379731.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009846.hg.1 |
ENST00000379731.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009846.hg.1 |
ENST00000379731.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009846.hg.1 |
ENST00000535206.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009846.hg.1 |
ENST00000535206.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009846.hg.1 |
ENST00000535206.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009846.hg.1 |
ENST00000535206.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009846.hg.1 |
NM_001497.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009846.hg.1 |
NM_001497.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009846.hg.1 |
NM_001497.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009846.hg.1 |
NM_001497.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009846.hg.1 |
NM_001497.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009846.hg.1 |
NM_001497.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009846.hg.1 |
NM_001497.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009846.hg.1 |
NM_001497.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900167368.hg.1 |
ENST00000379731.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900167368.hg.1 |
ENST00000535206.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900167368.hg.1 |
NM_001497.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900167368.hg.1 |
ENST00000379731.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900167368.hg.1 |
ENST00000535206.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900167368.hg.1 |
NM_001497.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900167368.hg.1 |
TC0900009846.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900167370.hg.1 |
ENST00000379731.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900167370.hg.1 |
ENST00000535206.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900167370.hg.1 |
NM_001497.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900167370.hg.1 |
ENST00000379731.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900167370.hg.1 |
ENST00000535206.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900167370.hg.1 |
NM_001497.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900167370.hg.1 |
TC0900009846.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900167361.hg.1 |
ENST00000535206.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900167361.hg.1 |
ENST00000535206.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900167361.hg.1 |
TC0900009846.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
ENST00000426270.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
ENST00000426270.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
ENST00000426270.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
ENST00000426270.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
ENST00000426270.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
ENST00000426270.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
ENST00000426270.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
ENST00000426270.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
ENST00000426270.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
ENST00000426270.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
ENST00000426270.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
ENST00000426270.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
ENST00000442432.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
ENST00000442432.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
ENST00000442432.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
ENST00000442432.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
ENST00000442432.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
ENST00000442432.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
ENST00000442432.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
ENST00000442432.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
ENST00000442432.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
NR_108108.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
NR_108108.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
NR_108108.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
NR_108108.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
NR_108108.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
NR_108108.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
NR_108108.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
NR_108108.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
NR_108108.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
NR_108108.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
NR_108108.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
NR_108109.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
NR_108109.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
NR_108109.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
NR_108109.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
NR_108109.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
NR_108109.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
NR_108109.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
NR_108109.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
NR_108109.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
NR_108109.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
NR_108109.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
NR_108110.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
NR_108110.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
NR_108110.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
NR_108110.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
NR_108110.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
NR_108110.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
NR_108110.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006966.hg.1 |
NR_108110.1 |
Refseq |