Assay Details
| Gene Symbol: NEMF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011537317.2 | XP_011535619.1 | 10 - 11 | 1174 | 144 | |
| XM_011537318.2 | XP_011535620.1 | 10 - 11 | 1174 | 144 | ||
| XM_017021760.1 | XP_016877249.1 | 9 - 10 | 877 | 144 | ||
| XM_017021761.1 | XP_016877250.1 | 4 - 5 | 330 | 144 | ||
| GenBank mRNA | AK000913.1 | - | 10 - 11 | 905 | 144 | |
| AK293236.1 | - | 9 - 10 | 789 | 144 | ||
| AK295010.1 | - | 9 - 10 | 839 | 144 | ||
| AK304695.1 | - | 10 - 11 | 910 | 144 | ||
| AK308537.1 | - | 8 - 9 | 687 | 144 | ||
| BC006001.1 | - | 9 - 10 | 791 | 144 | ||
| BC056687.1 | - | 10 - 11 | 921 | 144 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

NEMF
Genomic Map