Assay Details
| Gene Symbol: SLC26A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000111.2 | NP_000102.1 | 6 - 7 | 945 | 90 | |
| XM_011515867.2 | XP_011514169.1 | 6 - 7 | 926 | 90 | ||
| GenBank mRNA | BC025671.1 | - | 6 - 7 | 919 | 5202612 | 90 |
| BX640837.1 | - | 6 - 7 | 888 | 90 | ||
| L02785.1 | - | 6 - 7 | 918 | 90 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001746.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
ion transport
anion transport
excretion
bicarbonate transport
oxalate transport
regulation of membrane potential
sperm capacitation
regulation of intracellular pH
intracellular pH elevation
membrane hyperpolarization
cellular response to cAMP
sulfate transmembrane transport
chloride transmembrane transport
transcription factor activity, sequence-specific DNA binding
transcription cofactor activity
transporter activity
chloride channel activity
inorganic anion exchanger activity
protein binding
secondary active sulfate transmembrane transporter activity
bicarbonate transmembrane transporter activity
chloride transmembrane transporter activity
sulfate transmembrane transporter activity
anion:anion antiporter activity
oxalate transmembrane transporter activity
regulation of transcription, DNA-templated
ion transport
anion transport
excretion
bicarbonate transport
oxalate transport
regulation of membrane potential
sperm capacitation
regulation of intracellular pH
intracellular pH elevation
membrane hyperpolarization
cellular response to cAMP
sulfate transmembrane transport
chloride transmembrane transport
transcription factor activity, sequence-specific DNA binding
transcription cofactor activity
transporter activity
chloride channel activity
inorganic anion exchanger activity
protein binding
secondary active sulfate transmembrane transporter activity
bicarbonate transmembrane transporter activity
chloride transmembrane transporter activity
sulfate transmembrane transporter activity
anion:anion antiporter activity
oxalate transmembrane transporter activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07027153.hg.1 | TC07001746.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07027153.hg.1 | ENST00000340010.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07027153.hg.1 | ENST00000422236.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07027153.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000453332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000453332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000453332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000453332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000453332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000453332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000468551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000468551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000468551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000468551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000468551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000469651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000469651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000469651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | ENST00000469651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000453332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000453332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000453332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000453332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000453332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000453332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000468551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000468551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000468551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000468551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000468551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000469651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000469651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000469651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | ENST00000469651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012221.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700188105.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700188105.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700188105.hg.1 | ENST00000468551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700188105.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700188105.hg.1 | ENST00000340010.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700188105.hg.1 | ENST00000379083.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700188105.hg.1 | ENST00000468551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700188105.hg.1 | NM_000111.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700188105.hg.1 | TC0700012221.hg.1 | Clariom D TC |

SLC26A3
Genomic Map