Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MACRO domain containing 1 | MACROD1 | LRP16 | Sense | Chr. 11 - 63998554 - 64166652 on NCBI Build GRCh38 | |
| Downstream | stress induced phosphoprotein 1 | STIP1 |
HEL-S-94n, HOP, IEF-SSP-3521, P60, STI1, STI1L |
44.4kb | Anti-sense | Chr. 11 - 64185272 - 64204548 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map