Assay Details
| Gene Symbol: EML3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300793.1 | NP_001287722.1 | 21 - 22 | 2691 | 106 | |
| NM_001300794.1 | NP_001287723.1 | 21 - 22 | 2688 | 106 | ||
| XM_006718489.3 | XP_006718552.1 | 21 - 22 | 3063 | 106 | ||
| XM_011544896.2 | XP_011543198.1 | 21 - 22 | 3060 | 106 | ||
| XM_011544897.2 | XP_011543199.1 | 20 - 21 | 2907 | 106 | ||
| XM_017017480.1 | XP_016872969.1 | 21 - 22 | 2434 | 106 | ||
| XM_017017481.1 | XP_016872970.1 | 20 - 21 | 2320 | 106 | ||
| XM_017017483.1 | XP_016872972.1 | 21 - 22 | 2491 | 106 | ||
| GenBank mRNA | AK093146.1 | - | 20 - 21 | 2363 | 106 | |
| AK296901.1 | - | 21 - 22 | 2453 | 106 | ||
More Information
Set Membership: |
|

EML3
Genomic Map