Assay Details
| Gene Symbol: CDADC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193478.1 | NP_001180407.1 | 4 - 5 | 553 | 242 | |
| NM_030911.3 | NP_112173.1 | 4 - 5 | 553 | 242 | ||
| XM_011535250.2 | XP_011533552.1 | 4 - 5 | 517 | 242 | ||
| XM_011535251.1 | XP_011533553.1 | 4 - 5 | 497 | 242 | ||
| XM_011535253.1 | XP_011533555.1 | 4 - 5 | 449 | 102 | ||
| GenBank mRNA | AK312835.1 | - | 4 - 5 | 523 | 242 | |
| AY027525.1 | - | 4 - 5 | 502 | 242 | ||
| BC009562.1 | - | 4 - 5 | 377 | 102 | ||
| BC070238.1 | - | 4 - 5 | 521 | 242 | ||
| BC092434.1 | - | 4 - 5 | 531 | 242 | ||
| BC125202.1 | - | 4 - 5 | 467 | 40123430 | 242 | |
More Information
Set Membership: |
|

CDADC1
Genomic Map