Assay Details
| Gene Symbol: HSH2D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032855.3 | NP_116244.1 | 5 - 6 | 744 | 88 | |
| XM_011528386.2 | XP_011526688.1 | 3 - 4 | 339 | 88 | ||
| XM_011528388.2 | XP_011526690.1 | 4 - 5 | 419 | 88 | ||
| XM_011528390.1 | XP_011526692.1 | 3 - 4 | 330 | 88 | ||
| XM_017027402.1 | XP_016882891.1 | 3 - 4 | 330 | 88 | ||
| XM_017027403.1 | XP_016882892.1 | 3 - 4 | 330 | 88 | ||
| XM_017027404.1 | XP_016882893.1 | 3 - 4 | 340 | 88 | ||
| GenBank mRNA | AK027792.1 | - | 3 - 4 | 324 | 88 | |
| AY319652.1 | - | 5 - 6 | 744 | 88 | ||
| BC025237.2 | - | 2 - 3 | 244 | 4853087 | 88 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the XM_011528388.2 transcripts.
Set Membership: |
|

HSH2D
Genomic Map