| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05005517.hg.1 |
TC05000391.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05005517.hg.1 |
ENST00000296783.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05005517.hg.1 |
ENST00000423041.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05005517.hg.1 |
ENST00000428308.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05005517.hg.1 |
ENST00000453514.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05005517.hg.1 |
ENST00000504233.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05005517.hg.1 |
NM_001114393.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05005517.hg.1 |
NM_001114394.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05005517.hg.1 |
NM_173797.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05005518.hg.1 |
TC05000391.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05005518.hg.1 |
ENST00000296783.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05005518.hg.1 |
ENST00000423041.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05005518.hg.1 |
ENST00000428308.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05005518.hg.1 |
ENST00000453514.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05005518.hg.1 |
ENST00000504233.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05005518.hg.1 |
NM_001114393.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05005518.hg.1 |
NM_001114394.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05005518.hg.1 |
NM_173797.4 |
Refseq |