Assay Details
| Gene Symbol: ATP6AP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005765.2 | NP_005756.2 | 4 - 5 | 495 | 105 | |
| GenBank mRNA | AF109363.1 | - | 4 - 5 | 456 | 105 | |
| AF248966.1 | - | 4 - 5 | 500 | 105 | ||
| AF291814.1 | - | 4 - 5 | 482 | 105 | ||
| AK075382.1 | - | 4 - 5 | 553 | 105 | ||
| AK296614.1 | - | 3 - 4 | 358 | 105 | ||
| AK314633.1 | - | 4 - 5 | 446 | 105 | ||
| AL049929.1 | - | 3 - 4 | 335 | 105 | ||
| AY429341.1 | - | 3 - 4 | 358 | 105 | ||
| BC084541.1 | - | 4 - 5 | 475 | 6575732 | 105 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000190.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X002108.hg.1 | TC0X000190.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X002108.hg.1 | ENST00000378438.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X002108.hg.1 | ENST00000486558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X002108.hg.1 | ENST00000535539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X002108.hg.1 | ENST00000544975.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X002108.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000423649.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000423649.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000423649.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000423649.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000423649.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000423649.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000423649.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000423649.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000436783.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000436783.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000436783.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000436783.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000436783.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000436783.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000436783.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000447485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000447485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000447485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000447485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000447485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000447485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000447485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000447485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000479120.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000479120.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000479120.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000486558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000486558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000486558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000486558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000486558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000486558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000486558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000486558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000486558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000487051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000487051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000487051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00147684.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147684.hg.1 | ENST00000436783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147684.hg.1 | ENST00000447485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147684.hg.1 | ENST00000479120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147684.hg.1 | ENST00000486558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147684.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00147684.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147684.hg.1 | ENST00000436783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147684.hg.1 | ENST00000447485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147684.hg.1 | ENST00000479120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147684.hg.1 | ENST00000486558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147684.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00147684.hg.1 | TC0X00007012.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000423649.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000423649.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000423649.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000423649.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000423649.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000423649.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000423649.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000423649.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000436783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000436783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000436783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000436783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000436783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000436783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000436783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000447485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000447485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000447485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000447485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000447485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000447485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000447485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000447485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000479120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000479120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000479120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000486558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000486558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000486558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000486558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000486558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000486558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000486558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000486558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000486558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000487051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000487051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000487051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | NM_005765.2 | Refseq |

ATP6AP2
Genomic Map