Assay Details
| Gene Symbol: CALY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001321617.1 | NP_001308546.1 | 1 - 2 | 107 | ||
| NM_015722.3 | NP_056537.1 | 1 - 2 | 107 | |||
| GenBank mRNA | AF225903.1 | - | 1 - 2 | 70 | 108 | |
| BI914942.1 | - | 1 - 2 | 107 | |||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001787.hg
Additional Information:
This assay is in the 5'UTR region of the AF225903.1,NM_001321617.1,NM_015722.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10023614.hg.1 | ENST00000368556.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10023614.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10023614.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10023614.hg.1 | TC10001787.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000368555.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000368555.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000368555.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000368555.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000467433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000467433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000467433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000467433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000467433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000467433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000467611.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000467611.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000467611.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000467611.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000467611.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000467611.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000467611.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000605518.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10023610.hg.1 | ENST00000252939.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10023610.hg.1 | ENST00000368555.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10023610.hg.1 | ENST00000368556.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10023610.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10023610.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10023610.hg.1 | TC10001787.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10023612.hg.1 | ENST00000252939.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10023612.hg.1 | ENST00000368555.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10023612.hg.1 | ENST00000368556.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10023612.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10023612.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10023612.hg.1 | TC10001787.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10023613.hg.1 | ENST00000252939.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10023613.hg.1 | ENST00000368556.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10023613.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10023613.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10023613.hg.1 | TC10001787.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10023615.hg.1 | ENST00000368556.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10023615.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10023615.hg.1 | TC10001787.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1000184614.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184614.hg.1 | ENST00000368555.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184614.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184614.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000184614.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184614.hg.1 | ENST00000368555.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184614.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184614.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000184614.hg.1 | TC1000012395.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000184620.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000184620.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000184620.hg.1 | TC1000012395.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000184617.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184617.hg.1 | ENST00000368555.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184617.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184617.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000184617.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184617.hg.1 | ENST00000368555.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184617.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184617.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000184617.hg.1 | TC1000012395.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000184618.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184618.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184618.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000184618.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184618.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184618.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000184618.hg.1 | TC1000012395.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000184619.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184619.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000184619.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000184619.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000184619.hg.1 | TC1000012395.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000368555.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000368555.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000368555.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000368555.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000467433.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000467433.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000467433.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000467433.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000467433.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000467433.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000467611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000467611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000467611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000467611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000467611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000467611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000467611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000605518.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | NM_015722.3 | Refseq |

CALY
Genomic Map