Assay Details
| Gene Symbol: BTAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003972.2 | NP_003963.1 | 1 - 2 | 109 | 136 | |
| XM_011540326.1 | XP_011538628.1 | 1 - 2 | 319 | 136 | ||
| XM_011540329.2 | XP_011538631.1 | 1 - 2 | 319 | 136 | ||
| XM_017016877.1 | XP_016872366.1 | 1 - 2 | 318 | 136 | ||
| GenBank mRNA | AF038362.1 | - | 1 - 2 | 128 | 136 | |
| AK308399.1 | - | 1 - 2 | 318 | 136 | ||
| BC112201.1 | - | 1 - 2 | 93 | 8327669 | 136 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000655.hg
Additional Information:
This assay is in the 5'UTR region of the AF038362.1,BC112201.1,NM_003972.2,XM_011540326.1,XM_011540329.2,XM_017016877.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000265990.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000265990.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000265990.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000265990.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000265990.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000265990.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000265990.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000265990.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000265990.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000265990.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000265990.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000265990.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000265990.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000265990.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000265990.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000265990.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000265990.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000265990.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000265990.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000265990.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000265990.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000265990.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000265990.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000265990.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000265990.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000265990.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000265990.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000265990.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000265990.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000265990.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000265990.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000265990.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000265990.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000265990.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000265990.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000265990.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000265990.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000265990.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000265990.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000265990.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000265990.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000265990.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000265990.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000265990.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000471217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000471217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000471217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000471217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000471217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000471217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000471217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000471217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000471217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000471217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000471217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000471217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000471217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000471217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000471217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000471217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000476401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000476401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000476401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000476401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000544642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000544642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000544642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000544642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000544642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000544642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000544642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000544642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000544642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000544642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000544642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000544642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000544642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000544642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000544642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000544642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000544642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000544642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000544642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000544642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000544642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000544642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000544642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000544642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000544642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000544642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000544642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000544642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000544642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000544642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000544642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000544642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000544642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000544642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000544642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000544642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000544642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000544642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000544642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000544642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000544642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | ENST00000544642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | NM_003972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | NM_003972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | NM_003972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | NM_003972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | NM_003972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | NM_003972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | NM_003972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | NM_003972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | NM_003972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | NM_003972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | NM_003972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | NM_003972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | NM_003972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | NM_003972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | NM_003972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | NM_003972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | NM_003972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | NM_003972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | NM_003972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | NM_003972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | NM_003972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | NM_003972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | NM_003972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | NM_003972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | NM_003972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | NM_003972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | NM_003972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | NM_003972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | NM_003972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | NM_003972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | NM_003972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | NM_003972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | NM_003972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | NM_003972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | NM_003972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | NM_003972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | NM_003972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | NM_003972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | NM_003972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | NM_003972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | NM_003972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | NM_003972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008452.hg.1 | NM_003972.2 | Refseq |

BTAF1
Genomic Map