Assay Details
| Gene Symbol: NUB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016118.4 | NP_057202.3 | 12 - 13 | 1442 | 73 | |
| XM_017012305.1 | XP_016867794.1 | 12 - 13 | 1501 | 73 | ||
| XM_017012306.1 | XP_016867795.1 | 12 - 13 | 1523 | 73 | ||
| XM_017012313.1 | XP_016867802.1 | 13 - 14 | 1604 | 73 | ||
| XM_017012314.1 | XP_016867803.1 | 13 - 14 | 1480 | 73 | ||
| GenBank mRNA | AF155099.1 | - | 12 - 13 | 1374 | 73 | |
| AF300717.1 | - | 12 - 13 | 1431 | 73 | ||
| AY129295.1 | - | 12 - 13 | 1390 | 73 | ||
| BC046354.2 | - | 11 - 12 | 1358 | 6070128 | 73 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001016.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000413040.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000413040.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000413040.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000413040.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000413040.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000413040.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000413040.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000413040.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000413040.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000413040.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000413040.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000413040.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000413040.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000413040.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000413040.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000413040.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000413040.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000413040.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000413040.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000413040.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000413040.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000413040.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000413040.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000460712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000460712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000460712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000460712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000460712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000460712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000468404.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000468404.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000468404.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000468404.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000468404.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000468404.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000468404.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000468404.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000468404.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000468404.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000468404.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000468404.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000468404.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000470229.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000470229.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000470229.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000470229.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000470229.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000470229.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000470229.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000470316.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000470316.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000470316.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000470316.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000470316.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000470316.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000470316.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000470316.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000470316.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000470316.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000470316.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000470316.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000470316.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000477666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000477666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000477666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000477666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000477666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000477666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000477666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000480714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000480714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000480714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000480714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000480907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000480907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000480907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000480907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000480907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000480907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000480907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000480907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000480907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000480907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000480907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000483358.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000483358.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000483358.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000483358.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000483358.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000483358.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000483358.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000483358.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000493588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000493588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000493588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000493588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000493588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000493588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000493588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000493588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000493588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000493588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000497987.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000497987.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000497987.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000497987.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000497987.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000497987.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000497987.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000568733.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000568733.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000568733.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000568733.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000568733.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000568733.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000568733.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000568733.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000568733.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000568733.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000568733.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000568733.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000568733.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000568733.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000568733.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000568733.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000568733.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000568733.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000568733.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000568733.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000568733.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000568733.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000568733.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | ENST00000568733.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_001243351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_001243351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_001243351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_001243351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_001243351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_001243351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_001243351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_001243351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_001243351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_001243351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_001243351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_001243351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_001243351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_001243351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_001243351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_001243351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_001243351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_001243351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_001243351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_001243351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_001243351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_001243351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_001243351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_016118.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_016118.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_016118.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_016118.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_016118.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_016118.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_016118.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_016118.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_016118.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_016118.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_016118.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_016118.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_016118.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_016118.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_016118.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_016118.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_016118.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_016118.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_016118.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_016118.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_016118.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009700.hg.1 | NM_016118.4 | Refseq |

NUB1
Genomic Map