Assay Details
| Gene Symbol: WAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000377.2 | NP_000368.1 | 8 - 9 | 837 | 95 | |
| XM_011543977.2 | XP_011542279.1 | 8 - 9 | 842 | 95 | ||
| XM_017029786.1 | XP_016885275.1 | 8 - 9 | 881 | 95 | ||
| GenBank mRNA | BC002961.1 | - | 8 - 9 | 816 | 95 | |
| BC012738.2 | - | 8 - 9 | 808 | 3688197 | 95 | |
| U12707.1 | - | 8 - 9 | 814 | 95 | ||
| U19927.1 | - | 8 - 9 | 804 | 95 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000257.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of T cell antigen processing and presentation
protein complex assembly
defense response
immune response
blood coagulation
actin polymerization or depolymerization
epidermis development
endosomal transport
actin filament polymerization
actin filament-based movement
Fc-gamma receptor signaling pathway involved in phagocytosis
T cell activation
regulation of catalytic activity
T cell receptor signaling pathway
negative regulation of cell motility
positive regulation of Arp2/3 complex-mediated actin nucleation
actin binding
protein binding
SH3 domain binding
protein kinase binding
GTPase regulator activity
identical protein binding
phospholipase binding
regulation of T cell antigen processing and presentation
protein complex assembly
defense response
immune response
blood coagulation
actin polymerization or depolymerization
epidermis development
endosomal transport
actin filament polymerization
actin filament-based movement
Fc-gamma receptor signaling pathway involved in phagocytosis
T cell activation
regulation of catalytic activity
T cell receptor signaling pathway
negative regulation of cell motility
positive regulation of Arp2/3 complex-mediated actin nucleation
actin binding
protein binding
SH3 domain binding
protein kinase binding
GTPase regulator activity
identical protein binding
phospholipase binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X003168.hg.1 | TC0X000257.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X003168.hg.1 | ENST00000376701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X003168.hg.1 | NM_000377.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X003169.hg.1 | TC0X000257.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X003169.hg.1 | ENST00000376701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X003169.hg.1 | NM_000377.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000465982.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000465982.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000465982.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000465982.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000465982.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000465982.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000465982.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000465982.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000465982.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000465982.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000465982.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000470107.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000470107.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000470107.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000474174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000474174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000490627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000490627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000490627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000490627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000490627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00149223.hg.1 | ENST00000376701.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00149223.hg.1 | ENST00000474174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00149223.hg.1 | NM_000377.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00149223.hg.1 | ENST00000376701.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00149223.hg.1 | ENST00000474174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00149223.hg.1 | NM_000377.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00149223.hg.1 | TC0X00007199.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00149222.hg.1 | ENST00000376701.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00149222.hg.1 | ENST00000474174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00149222.hg.1 | NM_000377.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00149222.hg.1 | ENST00000376701.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00149222.hg.1 | ENST00000474174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00149222.hg.1 | NM_000377.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00149222.hg.1 | TC0X00007199.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000465982.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000465982.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000465982.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000465982.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000465982.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000465982.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000465982.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000465982.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000465982.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000465982.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000465982.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000470107.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000470107.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000470107.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000474174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000474174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000490627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000490627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000490627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000490627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000490627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | NM_000377.2 | Refseq |

WAS
Genomic Map