Assay Details
| Gene Symbol: H6PD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004285.3 | NP_004276.2 | 1 - 2 | 262 | 154 | |
| GenBank mRNA | AJ012590.1 | - | 1 - 2 | 261 | 154 | |
| BC081559.1 | - | 1 - 2 | 292 | 5266232 | 154 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000113.hg.1
Additional Information:
This assay is in the 5'UTR region of the AJ012590.1,BC081559.1,NM_004285.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01001734.hg.1 | TC01000113.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001734.hg.1 | ENST00000377403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001734.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001735.hg.1 | TC01000113.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001735.hg.1 | ENST00000377403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001735.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000495451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000495451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000495451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000495451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000602477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000602477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000602477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000602477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000602477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000602477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000602477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000602477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146831.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146831.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146831.hg.1 | TC0100006771.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100146836.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146836.hg.1 | ENST00000602477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146836.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146836.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146836.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146836.hg.1 | ENST00000602477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146836.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146836.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146836.hg.1 | TC0100006771.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000495451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000495451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000495451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000495451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000602477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000602477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000602477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000602477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000602477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000602477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000602477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000602477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146832.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146832.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146832.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146832.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146832.hg.1 | TC0100006771.hg.1 | Clariom D TC |

H6PD
Genomic Map