Assay Details
| Gene Symbol: H6PD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282587.1 | NP_001269516.1 | 4 - 5 | 1203 | 81 | |
| NM_004285.3 | NP_004276.2 | 4 - 5 | 1291 | 81 | ||
| XM_005263540.4 | XP_005263597.1 | 4 - 5 | 3967 | 81 | ||
| XM_006711052.3 | XP_006711115.1 | 4 - 5 | 1320 | 81 | ||
| XM_017002865.1 | XP_016858354.1 | 4 - 5 | 3574 | 81 | ||
| GenBank mRNA | BC081559.1 | - | 4 - 5 | 1321 | 5266232 | 81 |
| CR749282.1 | - | 4 - 5 | 1203 | 81 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000113.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01001738.hg.1 | TC01000113.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001738.hg.1 | ENST00000377403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001738.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001741.hg.1 | TC01000113.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001741.hg.1 | ENST00000377403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001741.hg.1 | ENST00000495451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001741.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001744.hg.1 | TC01000113.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001744.hg.1 | ENST00000377403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001744.hg.1 | ENST00000495451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001744.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001743.hg.1 | TC01000113.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001743.hg.1 | ENST00000377403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001743.hg.1 | ENST00000495451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001743.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000495451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000495451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000495451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000495451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000602477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000602477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000602477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000602477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000602477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000602477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000602477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | ENST00000602477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006771.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001740.hg.1 | TC01000113.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001740.hg.1 | ENST00000377403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001740.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146841.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146841.hg.1 | ENST00000495451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146841.hg.1 | ENST00000602477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146841.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146841.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146841.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146841.hg.1 | ENST00000495451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146841.hg.1 | ENST00000602477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146841.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146841.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146841.hg.1 | TC0100006771.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100146839.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146839.hg.1 | ENST00000602477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146839.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146839.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146839.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146839.hg.1 | ENST00000602477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146839.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146839.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146839.hg.1 | TC0100006771.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100146840.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146840.hg.1 | ENST00000602477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146840.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146840.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146840.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146840.hg.1 | ENST00000602477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146840.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146840.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146840.hg.1 | TC0100006771.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100146842.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146842.hg.1 | ENST00000495451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146842.hg.1 | ENST00000602477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146842.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146842.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146842.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146842.hg.1 | ENST00000495451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146842.hg.1 | ENST00000602477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146842.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146842.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146842.hg.1 | TC0100006771.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000377403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000495451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000495451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000495451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000495451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000602477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000602477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000602477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000602477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000602477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000602477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000602477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | ENST00000602477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | NM_001282587.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | NM_004285.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006771.hg.1 | NM_004285.3 | Refseq |

H6PD
Genomic Map