Assay Details
| Gene Symbol: MIDN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177401.4 | NP_796375.3 | 1 - 2 | 107 | 72 | |
| XM_005259671.3 | XP_005259728.1 | 1 - 2 | 107 | 72 | ||
| XM_005259672.3 | XP_005259729.1 | 1 - 2 | 108 | 72 | ||
| GenBank mRNA | BM966744.1 | - | 1 - 2 | 107 | 72 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000033.hg
Additional Information:
This assay is in the 5'UTR region of the NM_177401.4,XM_005259671.3,XM_005259672.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000586843.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000586843.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000586843.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000586843.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000586843.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000590136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000590136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000590136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000590136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19000414.hg.1 | ENST00000300952.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19000414.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19000414.hg.1 | TC19000033.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19000416.hg.1 | ENST00000300952.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19000416.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19000416.hg.1 | TC19000033.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000586843.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000586843.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000586843.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000586843.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000586843.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000590136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000590136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000590136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000590136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900145030.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900145030.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900145030.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900145030.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900145030.hg.1 | TC1900006522.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900145030.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900145030.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900145030.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900145030.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900145028.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900145028.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900145028.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900145028.hg.1 | TC1900006522.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900145028.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900145028.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900145028.hg.1 | NM_177401.4 | Refseq |

MIDN
Genomic Map