Assay Details
| Gene Symbol: MIDN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177401.4 | NP_796375.3 | 5 - 6 | 1214 | 157 | |
| XM_005259671.3 | XP_005259728.1 | 6 - 7 | 1343 | 157 | ||
| GenBank mRNA | BC094778.1 | - | 4 - 5 | 1108 | 30342357 | 157 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000033.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19000420.hg.1 | ENST00000300952.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19000420.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19000420.hg.1 | TC19000033.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000586843.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000586843.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000586843.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000586843.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000586843.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000590136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000590136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000590136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000590136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900145042.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900145042.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900145042.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900145042.hg.1 | TC1900006522.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900145042.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900145042.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900145042.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900145041.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900145041.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900145041.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900145041.hg.1 | TC1900006522.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900145041.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900145041.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900145041.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000300952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000586757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000586843.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000586843.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000586843.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000586843.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000586843.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000590136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000590136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000590136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000590136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | ENST00000591446.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006522.hg.1 | NM_177401.4 | Refseq |

MIDN
Genomic Map