Assay Details
| Gene Symbol: CAMTA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015215.3 | NP_056030.1 | 19 - 20 | 4893 | 83 | |
| XM_011541086.2 | XP_011539388.1 | 19 - 20 | 4757 | 83 | ||
| XM_017000775.1 | XP_016856264.1 | 18 - 19 | 4687 | 83 | ||
| XM_017000777.1 | XP_016856266.1 | 19 - 20 | 4418 | 83 | ||
| XM_017000778.1 | XP_016856267.1 | 19 - 20 | 4418 | 83 | ||
| GenBank mRNA | AB020640.3 | - | 19 - 20 | 4893 | 83 | |
| AF111804.1 | - | 3 - 4 | 383 | 83 | ||
| AK302088.1 | - | 12 - 13 | 1887 | 83 | ||
| AK309727.1 | - | 9 - 10 | 1900 | 83 | ||
| AY349360.1 | - | 3 - 4 | 378 | 83 | ||
| BC151835.1 | - | 19 - 20 | 4893 | 8860115 | 83 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000096.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase II promoter
neuromuscular process controlling balance
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
sequence-specific DNA binding
transcription from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase II promoter
neuromuscular process controlling balance
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
sequence-specific DNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000303635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000303635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000303635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000303635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000303635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000303635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000303635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000303635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000303635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000303635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000303635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000303635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000303635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000303635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000303635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000303635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000303635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000303635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000303635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000303635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000303635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000303635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000303635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000303635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000303635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000303635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000303635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000303635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000303635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000303635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000303635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000461311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000461311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000461311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000461311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000461311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000461580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000461580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000461580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000461580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000461580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000461580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000461580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000461580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000467267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000467267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000467267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000467267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000467267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000467404.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000467404.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000467404.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000467404.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000470648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000470648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000470648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000470648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000470648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000470648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000470648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000470648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000472283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000472283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000472283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000473578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000473578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000473578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000473578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000473578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000476163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000476163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000476163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000476163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000476163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000476163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000476163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000476163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000476864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000476864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000476864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000476864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000476864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000476864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000476864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000476864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000482934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000482934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000482934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000482934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000482934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000482934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000482934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000486138.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000486138.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000486138.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000486138.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000490738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000490738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000490738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000490738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000490905.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000490905.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000490905.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000490905.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000490905.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000490905.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000490905.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000490905.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000490905.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000495233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000495233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000495233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000495233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000495233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000495233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000495233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000495233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000495233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000495233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000495233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000495233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000495233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000495233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000495233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000557126.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000557126.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000557126.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000557126.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000557126.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | ENST00000557126.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_001195563.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_001195563.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_001195563.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_001195563.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_001195563.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_001195563.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_001242701.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_001242701.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_001242701.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_001242701.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_001242701.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_001242701.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_001242701.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_001242701.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_001242701.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_015215.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_015215.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_015215.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_015215.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_015215.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_015215.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_015215.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_015215.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_015215.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_015215.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_015215.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_015215.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_015215.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_015215.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_015215.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_015215.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_015215.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_015215.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_015215.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_015215.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_015215.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_015215.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_015215.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_015215.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_015215.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_015215.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_015215.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_015215.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_015215.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_015215.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NM_015215.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NR_038934.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NR_038934.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NR_038934.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NR_038934.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NR_038934.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NR_038934.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NR_038934.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NR_038934.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006707.hg.1 | NR_038934.1 | Refseq |

CAMTA1
Genomic Map