| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20006968.hg.1 |
ENST00000370487.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20006968.hg.1 |
NM_018270.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20006968.hg.1 |
TC20000497.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20006971.hg.1 |
ENST00000370487.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20006971.hg.1 |
NM_018270.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20006971.hg.1 |
TC20000497.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20006970.hg.1 |
ENST00000370487.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20006970.hg.1 |
NM_018270.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20006970.hg.1 |
TC20000497.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008029.hg.1 |
ENST00000370487.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008029.hg.1 |
ENST00000370487.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008029.hg.1 |
ENST00000370487.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008029.hg.1 |
ENST00000370487.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008029.hg.1 |
ENST00000370487.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008029.hg.1 |
ENST00000370487.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008029.hg.1 |
ENST00000370487.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008029.hg.1 |
ENST00000370487.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008029.hg.1 |
ENST00000370487.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008029.hg.1 |
ENST00000370487.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008029.hg.1 |
ENST00000370487.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008029.hg.1 |
NM_018270.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008029.hg.1 |
NM_018270.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008029.hg.1 |
NM_018270.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008029.hg.1 |
NM_018270.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008029.hg.1 |
NM_018270.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008029.hg.1 |
NM_018270.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008029.hg.1 |
NM_018270.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008029.hg.1 |
NM_018270.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000155883.hg.1 |
ENST00000370487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000155883.hg.1 |
NM_018270.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000155883.hg.1 |
TC2000008029.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000155883.hg.1 |
ENST00000370487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000155883.hg.1 |
NM_018270.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008029.hg.1 |
ENST00000370487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008029.hg.1 |
ENST00000370487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008029.hg.1 |
ENST00000370487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008029.hg.1 |
ENST00000370487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008029.hg.1 |
ENST00000370487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008029.hg.1 |
ENST00000370487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008029.hg.1 |
ENST00000370487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008029.hg.1 |
ENST00000370487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008029.hg.1 |
ENST00000370487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008029.hg.1 |
ENST00000370487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008029.hg.1 |
ENST00000370487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008029.hg.1 |
NM_018270.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008029.hg.1 |
NM_018270.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008029.hg.1 |
NM_018270.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008029.hg.1 |
NM_018270.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008029.hg.1 |
NM_018270.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008029.hg.1 |
NM_018270.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008029.hg.1 |
NM_018270.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008029.hg.1 |
NM_018270.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000155887.hg.1 |
ENST00000370487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000155887.hg.1 |
NM_018270.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000155887.hg.1 |
TC2000008029.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000155887.hg.1 |
ENST00000370487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000155887.hg.1 |
NM_018270.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000155886.hg.1 |
ENST00000370487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000155886.hg.1 |
NM_018270.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000155886.hg.1 |
TC2000008029.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000155886.hg.1 |
ENST00000370487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000155886.hg.1 |
NM_018270.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000155885.hg.1 |
ENST00000370487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000155885.hg.1 |
NM_018270.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000155885.hg.1 |
TC2000008029.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000155885.hg.1 |
ENST00000370487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000155885.hg.1 |
NM_018270.5 |
Refseq |