Assay Details
| Gene Symbol: ZBTB32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014383.2 | NP_055198.1 | 3 - 4 | 1238 | 125 | |
| XM_011526718.2 | XP_011525020.1 | 2 - 3 | 1399 | 125 | ||
| XM_017026588.1 | XP_016882077.1 | 1 - 2 | 904 | 125 | ||
| XM_017026589.1 | XP_016882078.1 | 3 - 4 | 1609 | 125 | ||
| GenBank mRNA | AF130255.1 | - | 2 - 3 | 1090 | 125 | |
| AF165097.1 | - | 1 - 2 | 880 | 125 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000481.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19006329.hg.1 | ENST00000262630.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19006329.hg.1 | ENST00000392197.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19006329.hg.1 | NM_014383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19006329.hg.1 | TC19000481.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19006330.hg.1 | ENST00000262630.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19006330.hg.1 | ENST00000392197.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19006330.hg.1 | NM_014383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19006330.hg.1 | TC19000481.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | ENST00000262630.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | ENST00000262630.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | ENST00000262630.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | ENST00000262630.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | ENST00000262630.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | ENST00000262630.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | ENST00000262630.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | ENST00000262630.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | ENST00000262630.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | ENST00000262630.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | ENST00000392197.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | ENST00000392197.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | ENST00000392197.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | ENST00000392197.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | ENST00000392197.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | ENST00000392197.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | ENST00000392197.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | ENST00000392197.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | ENST00000392197.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | ENST00000392197.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | ENST00000392197.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | ENST00000392197.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | ENST00000426659.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | ENST00000426659.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | ENST00000426659.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | ENST00000426659.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | ENST00000426659.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | ENST00000442282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | ENST00000442282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | ENST00000442282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | ENST00000442282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | ENST00000442282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | ENST00000451726.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | ENST00000451726.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | ENST00000451726.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | ENST00000481182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | ENST00000481182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | ENST00000481182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | ENST00000481182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | ENST00000481182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | ENST00000481182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | ENST00000481182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | NM_014383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | NM_014383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | NM_014383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | NM_014383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | NM_014383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | NM_014383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | NM_014383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | NM_014383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | NM_014383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007876.hg.1 | NM_014383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19006328.hg.1 | ENST00000262630.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19006328.hg.1 | ENST00000392197.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19006328.hg.1 | NM_014383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19006328.hg.1 | TC19000481.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1900158469.hg.1 | ENST00000262630.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900158469.hg.1 | ENST00000392197.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900158469.hg.1 | ENST00000426659.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900158469.hg.1 | ENST00000481182.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900158469.hg.1 | NM_014383.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900158469.hg.1 | TC1900007876.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900158469.hg.1 | ENST00000262630.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900158469.hg.1 | ENST00000392197.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900158469.hg.1 | ENST00000426659.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900158469.hg.1 | ENST00000481182.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900158469.hg.1 | NM_014383.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900158466.hg.1 | ENST00000262630.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900158466.hg.1 | ENST00000392197.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900158466.hg.1 | ENST00000426659.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900158466.hg.1 | ENST00000481182.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900158466.hg.1 | NM_014383.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900158466.hg.1 | TC1900007876.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900158466.hg.1 | ENST00000262630.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900158466.hg.1 | ENST00000392197.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900158466.hg.1 | ENST00000426659.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900158466.hg.1 | ENST00000481182.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900158466.hg.1 | NM_014383.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900158467.hg.1 | ENST00000262630.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900158467.hg.1 | ENST00000392197.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900158467.hg.1 | NM_014383.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900158467.hg.1 | TC1900007876.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900158467.hg.1 | ENST00000262630.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900158467.hg.1 | ENST00000392197.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900158467.hg.1 | NM_014383.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900158468.hg.1 | ENST00000262630.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900158468.hg.1 | ENST00000392197.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900158468.hg.1 | ENST00000426659.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900158468.hg.1 | NM_014383.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900158468.hg.1 | TC1900007876.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900158468.hg.1 | ENST00000262630.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900158468.hg.1 | ENST00000392197.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900158468.hg.1 | ENST00000426659.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900158468.hg.1 | NM_014383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | ENST00000262630.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | ENST00000262630.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | ENST00000262630.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | ENST00000262630.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | ENST00000262630.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | ENST00000262630.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | ENST00000262630.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | ENST00000262630.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | ENST00000262630.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | ENST00000262630.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | ENST00000392197.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | ENST00000392197.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | ENST00000392197.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | ENST00000392197.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | ENST00000392197.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | ENST00000392197.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | ENST00000392197.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | ENST00000392197.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | ENST00000392197.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | ENST00000392197.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | ENST00000392197.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | ENST00000392197.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | ENST00000426659.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | ENST00000426659.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | ENST00000426659.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | ENST00000426659.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | ENST00000426659.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | ENST00000442282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | ENST00000442282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | ENST00000442282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | ENST00000442282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | ENST00000442282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | ENST00000451726.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | ENST00000451726.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | ENST00000451726.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | ENST00000481182.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | ENST00000481182.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | ENST00000481182.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | ENST00000481182.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | ENST00000481182.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | ENST00000481182.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | ENST00000481182.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | NM_014383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | NM_014383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | NM_014383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | NM_014383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | NM_014383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | NM_014383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | NM_014383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | NM_014383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | NM_014383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007876.hg.1 | NM_014383.2 | Refseq |

ZBTB32
Genomic Map