Assay Details
| Gene Symbol: PARVA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018222.4 | NP_060692.2 | 3 - 4 | 461 | 125 | |
| XM_005253015.3 | XP_005253072.1 | 3 - 4 | 450 | 125 | ||
| GenBank mRNA | AF237771.1 | - | 3 - 4 | 384 | 125 | |
| AF325830.1 | - | 3 - 4 | 292 | 125 | ||
| AK001655.1 | - | 3 - 4 | 341 | 125 | ||
| AK022316.1 | - | 3 - 4 | 377 | 125 | ||
| AK098504.1 | - | 2 - 3 | 385 | 125 | ||
| AK304444.1 | - | 3 - 4 | 269 | 125 | ||
| BC014535.1 | - | 3 - 4 | 320 | 4065820 | 125 | |
| BC016713.1 | - | 3 - 4 | 378 | 4065758 | 125 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000201.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
sprouting angiogenesis
outflow tract septum morphogenesis
establishment or maintenance of cell polarity
regulation of cell shape
actin cytoskeleton reorganization
heterotypic cell-cell adhesion
substrate adhesion-dependent cell spreading
cilium morphogenesis
actin-mediated cell contraction
smooth muscle cell chemotaxis
actin binding
protein binding
cadherin binding involved in cell-cell adhesion
sprouting angiogenesis
outflow tract septum morphogenesis
establishment or maintenance of cell polarity
regulation of cell shape
actin cytoskeleton reorganization
heterotypic cell-cell adhesion
substrate adhesion-dependent cell spreading
cilium morphogenesis
actin-mediated cell contraction
smooth muscle cell chemotaxis
actin binding
protein binding
cadherin binding involved in cell-cell adhesion
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11002703.hg.1 | ENST00000334956.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11002703.hg.1 | ENST00000526746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11002703.hg.1 | ENST00000538608.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11002703.hg.1 | ENST00000539723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11002703.hg.1 | ENST00000550549.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11002703.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11002703.hg.1 | TC11000201.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000526746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000526746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000526746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000526746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000530755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000530755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000530755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000530755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000530755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000530755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000530755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533392.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533392.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533392.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533392.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000526746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000526746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000526746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000526746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000530755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000530755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000530755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000530755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000530755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000530755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000530755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533392.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533392.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533392.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533392.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100148444.hg.1 | ENST00000334956.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100148444.hg.1 | ENST00000526746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100148444.hg.1 | ENST00000528916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100148444.hg.1 | ENST00000530755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100148444.hg.1 | ENST00000533345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100148444.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100148444.hg.1 | ENST00000334956.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100148444.hg.1 | ENST00000526746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100148444.hg.1 | ENST00000528916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100148444.hg.1 | ENST00000530755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100148444.hg.1 | ENST00000533345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100148444.hg.1 | NM_018222.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100148444.hg.1 | TC1100006879.hg.1 | Clariom D TC |

PARVA
Genomic Map