Assay Details
| Gene Symbol: WDR20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242415.1 | NP_001229344.1 | 1 - 2 | 325 | 103 | |
| NM_001242417.1 | NP_001229346.1 | 1 - 2 | 325 | 103 | ||
| NM_001320130.1 | NP_001307059.1 | 1 - 2 | 325 | 103 | ||
| NM_144574.3 | NP_653175.2 | 1 - 2 | 325 | 103 | ||
| NM_181291.2 | NP_851808.1 | 1 - 2 | 325 | 103 | ||
| XM_006720318.2 | XP_006720381.1 | 1 - 2 | 275 | 103 | ||
| XM_011537338.1 | XP_011535640.1 | 1 - 2 | 291 | 103 | ||
| XM_017021766.1 | XP_016877255.1 | 1 - 2 | 291 | 103 | ||
| XM_017021768.1 | XP_016877257.1 | 1 - 2 | 358 | 103 | ||
| XM_017021770.1 | XP_016877259.1 | 1 - 2 | 358 | 103 | ||
| XM_017021781.1 | XP_016877270.1 | 1 - 2 | 275 | 103 | ||
| XM_017021783.1 | XP_016877272.1 | 1 - 2 | 291 | 103 | ||
| XM_017021784.1 | XP_016877273.1 | 1 - 2 | 358 | 103 | ||
| XM_017021785.1 | XP_016877274.1 | 1 - 2 | 291 | 103 | ||
| GenBank mRNA | AF327354.1 | - | 1 - 2 | 250 | 103 | |
| AK297727.1 | - | 1 - 2 | 263 | 103 | ||
| BC028387.2 | - | 1 - 2 | 284 | 4823662 | 103 | |
| BC030654.2 | - | 1 - 2 | 304 | 103 | ||
| BM707311.1 | - | 1 - 2 | 229 | 103 | ||
| BP363522.1 | - | 1 - 2 | 325 | 103 | ||
| DR423616.1 | - | 1 - 2 | 247 | 103 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017021781.1 transcripts.
Set Membership: |
|

WDR20
Genomic Map