Assay Details
| Gene Symbol: TPH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173353.3 | NP_775489.2 | 8 - 9 | 1216 | 78 | |
| GenBank mRNA | AY098914.1 | - | 8 - 9 | 1216 | 78 | |
| BC114442.1 | - | 8 - 9 | 1171 | 40083878 | 78 | |
| BC114499.1 | - | 8 - 9 | 1153 | 40083876 | 78 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000629.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
circadian rhythm
aromatic amino acid family metabolic process
response to activity
response to nutrient levels
serotonin biosynthetic process
response to estrogen
indolalkylamine biosynthetic process
response to glucocorticoid
response to calcium ion
oxidation-reduction process
cellular response to lithium ion
tryptophan 5-monooxygenase activity
iron ion binding
amino acid binding
circadian rhythm
aromatic amino acid family metabolic process
response to activity
response to nutrient levels
serotonin biosynthetic process
response to estrogen
indolalkylamine biosynthetic process
response to glucocorticoid
response to calcium ion
oxidation-reduction process
cellular response to lithium ion
tryptophan 5-monooxygenase activity
iron ion binding
amino acid binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | ENST00000333850.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | ENST00000333850.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | ENST00000333850.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | ENST00000333850.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | ENST00000333850.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | ENST00000333850.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | ENST00000333850.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | ENST00000333850.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | ENST00000333850.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | ENST00000333850.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | ENST00000333850.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | ENST00000333850.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | ENST00000333850.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | ENST00000333850.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | ENST00000333850.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | ENST00000546576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | ENST00000546576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | ENST00000546576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | ENST00000546576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | ENST00000546576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | ENST00000546576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | ENST00000546576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | ENST00000547278.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | ENST00000547278.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | ENST00000547278.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | ENST00000547278.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | ENST00000547278.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | ENST00000547278.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | ENST00000547348.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | ENST00000547348.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | ENST00000547348.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | ENST00000547348.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | ENST00000550403.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | ENST00000550403.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | ENST00000550403.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | ENST00000550403.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | ENST00000550403.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | ENST00000550403.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | ENST00000551074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | ENST00000551074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | ENST00000551074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | ENST00000551074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | NM_173353.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | NM_173353.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | NM_173353.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | NM_173353.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | NM_173353.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | NM_173353.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | NM_173353.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | NM_173353.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | NM_173353.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | NM_173353.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | NM_173353.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | NM_173353.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | NM_173353.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | NM_173353.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012667.hg.1 | NM_173353.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12008418.hg.1 | ENST00000333850.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12008418.hg.1 | NM_173353.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12008418.hg.1 | TC12000629.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | ENST00000333850.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | ENST00000333850.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | ENST00000333850.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | ENST00000333850.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | ENST00000333850.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | ENST00000333850.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | ENST00000333850.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | ENST00000333850.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | ENST00000333850.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | ENST00000333850.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | ENST00000333850.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | ENST00000333850.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | ENST00000333850.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | ENST00000333850.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | ENST00000333850.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | ENST00000546576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | ENST00000546576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | ENST00000546576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | ENST00000546576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | ENST00000546576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | ENST00000546576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | ENST00000546576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | ENST00000547278.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | ENST00000547278.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | ENST00000547278.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | ENST00000547278.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | ENST00000547278.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | ENST00000547278.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | ENST00000547348.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | ENST00000547348.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | ENST00000547348.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | ENST00000547348.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | ENST00000550403.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | ENST00000550403.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | ENST00000550403.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | ENST00000550403.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | ENST00000550403.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | ENST00000550403.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | ENST00000551074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | ENST00000551074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | ENST00000551074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | ENST00000551074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | NM_173353.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | NM_173353.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | NM_173353.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | NM_173353.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | NM_173353.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | NM_173353.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | NM_173353.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | NM_173353.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | NM_173353.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | NM_173353.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | NM_173353.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | NM_173353.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | NM_173353.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | NM_173353.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012667.hg.1 | NM_173353.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200201071.hg.1 | ENST00000333850.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201071.hg.1 | NM_173353.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200201071.hg.1 | ENST00000333850.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201071.hg.1 | NM_173353.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200201071.hg.1 | TC1200012667.hg.1 | Clariom D TC |

TPH2
Genomic Map