Assay Details
| Gene Symbol: PGRMC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282621.1 | NP_001269550.1 | 1 - 2 | 442 | 58 | |
| GenBank mRNA | AK293618.1 | - | 1 - 2 | 362 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008240.hg.1 | ENST00000217971.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008240.hg.1 | ENST00000217971.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008240.hg.1 | ENST00000217971.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008240.hg.1 | ENST00000217971.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008240.hg.1 | ENST00000217971.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008240.hg.1 | ENST00000217971.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008240.hg.1 | ENST00000535419.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008240.hg.1 | ENST00000535419.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008240.hg.1 | ENST00000535419.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008240.hg.1 | ENST00000535419.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008240.hg.1 | ENST00000535419.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008240.hg.1 | NM_001282621.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008240.hg.1 | NM_001282621.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008240.hg.1 | NM_001282621.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008240.hg.1 | NM_001282621.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008240.hg.1 | NM_001282621.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008240.hg.1 | NM_006667.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008240.hg.1 | NM_006667.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008240.hg.1 | NM_006667.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008240.hg.1 | NM_006667.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008240.hg.1 | NM_006667.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008240.hg.1 | NM_006667.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00156306.hg.1 | ENST00000217971.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00156306.hg.1 | ENST00000535419.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00156306.hg.1 | NM_001282621.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00156306.hg.1 | NM_006667.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00156306.hg.1 | ENST00000217971.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00156306.hg.1 | ENST00000535419.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00156306.hg.1 | NM_001282621.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00156306.hg.1 | NM_006667.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00156306.hg.1 | TC0X00008240.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00008240.hg.1 | ENST00000217971.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008240.hg.1 | ENST00000217971.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008240.hg.1 | ENST00000217971.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008240.hg.1 | ENST00000217971.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008240.hg.1 | ENST00000217971.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008240.hg.1 | ENST00000217971.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008240.hg.1 | ENST00000535419.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008240.hg.1 | ENST00000535419.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008240.hg.1 | ENST00000535419.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008240.hg.1 | ENST00000535419.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008240.hg.1 | ENST00000535419.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008240.hg.1 | NM_001282621.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008240.hg.1 | NM_001282621.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008240.hg.1 | NM_001282621.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008240.hg.1 | NM_001282621.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008240.hg.1 | NM_001282621.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008240.hg.1 | NM_006667.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008240.hg.1 | NM_006667.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008240.hg.1 | NM_006667.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008240.hg.1 | NM_006667.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008240.hg.1 | NM_006667.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008240.hg.1 | NM_006667.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00156307.hg.1 | ENST00000217971.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00156307.hg.1 | ENST00000535419.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00156307.hg.1 | NM_001282621.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00156307.hg.1 | NM_006667.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00156307.hg.1 | ENST00000217971.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00156307.hg.1 | ENST00000535419.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00156307.hg.1 | NM_001282621.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00156307.hg.1 | NM_006667.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00156307.hg.1 | TC0X00008240.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00156309.hg.1 | ENST00000217971.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00156309.hg.1 | ENST00000535419.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00156309.hg.1 | NM_001282621.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00156309.hg.1 | NM_006667.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00156309.hg.1 | ENST00000217971.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00156309.hg.1 | ENST00000535419.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00156309.hg.1 | NM_001282621.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00156309.hg.1 | NM_006667.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00156309.hg.1 | TC0X00008240.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00156305.hg.1 | ENST00000217971.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00156305.hg.1 | ENST00000535419.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00156305.hg.1 | NM_001282621.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00156305.hg.1 | NM_006667.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00156305.hg.1 | ENST00000217971.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00156305.hg.1 | ENST00000535419.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00156305.hg.1 | NM_001282621.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00156305.hg.1 | NM_006667.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00156305.hg.1 | TC0X00008240.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00156310.hg.1 | ENST00000217971.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00156310.hg.1 | ENST00000535419.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00156310.hg.1 | NM_001282621.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00156310.hg.1 | NM_006667.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00156310.hg.1 | ENST00000217971.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00156310.hg.1 | ENST00000535419.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00156310.hg.1 | NM_001282621.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00156310.hg.1 | NM_006667.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00156310.hg.1 | TC0X00008240.hg.1 | Clariom D TC |

PGRMC1
Genomic Map