Assay Details
| Gene Symbol: KRT36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003771.4 | NP_003762.1 | 3 - 4 | 694 | 61 | |
| GenBank mRNA | BC043581.1 | - | 4 - 5 | 757 | 5227645 | 61 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001507.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17019852.hg.1 | ENST00000328119.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019852.hg.1 | ENST00000393986.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019852.hg.1 | NM_003771.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019852.hg.1 | TC17001507.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010674.hg.1 | ENST00000328119.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010674.hg.1 | ENST00000328119.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010674.hg.1 | ENST00000328119.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010674.hg.1 | ENST00000328119.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010674.hg.1 | ENST00000328119.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010674.hg.1 | ENST00000328119.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010674.hg.1 | ENST00000328119.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010674.hg.1 | ENST00000328119.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010674.hg.1 | ENST00000328119.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010674.hg.1 | ENST00000393986.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010674.hg.1 | ENST00000393986.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010674.hg.1 | ENST00000393986.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010674.hg.1 | ENST00000393986.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010674.hg.1 | ENST00000393986.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010674.hg.1 | ENST00000393986.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010674.hg.1 | ENST00000393986.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010674.hg.1 | ENST00000393986.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010674.hg.1 | ENST00000393986.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010674.hg.1 | NM_003771.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010674.hg.1 | NM_003771.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010674.hg.1 | NM_003771.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010674.hg.1 | NM_003771.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010674.hg.1 | NM_003771.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010674.hg.1 | NM_003771.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010674.hg.1 | NM_003771.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010674.hg.1 | NM_003771.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010674.hg.1 | NM_003771.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019854.hg.1 | ENST00000328119.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019854.hg.1 | ENST00000393986.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019854.hg.1 | NM_003771.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019854.hg.1 | TC17001507.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019853.hg.1 | ENST00000328119.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019853.hg.1 | ENST00000393986.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019853.hg.1 | NM_003771.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019853.hg.1 | TC17001507.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019855.hg.1 | ENST00000328119.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019855.hg.1 | ENST00000393986.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019855.hg.1 | NM_003771.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019855.hg.1 | TC17001507.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1700184359.hg.1 | ENST00000328119.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184359.hg.1 | ENST00000393986.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184359.hg.1 | NM_003771.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700184359.hg.1 | ENST00000328119.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184359.hg.1 | ENST00000393986.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184359.hg.1 | NM_003771.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700184359.hg.1 | TC1700010674.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700184362.hg.1 | ENST00000328119.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184362.hg.1 | ENST00000393986.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184362.hg.1 | NM_003771.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700184362.hg.1 | ENST00000328119.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184362.hg.1 | ENST00000393986.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184362.hg.1 | NM_003771.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700184362.hg.1 | TC1700010674.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700184361.hg.1 | ENST00000328119.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184361.hg.1 | ENST00000393986.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184361.hg.1 | NM_003771.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700184361.hg.1 | ENST00000328119.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184361.hg.1 | ENST00000393986.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184361.hg.1 | NM_003771.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700184361.hg.1 | TC1700010674.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700184365.hg.1 | ENST00000393986.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184365.hg.1 | ENST00000393986.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184365.hg.1 | TC1700010674.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700010674.hg.1 | ENST00000328119.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010674.hg.1 | ENST00000328119.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010674.hg.1 | ENST00000328119.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010674.hg.1 | ENST00000328119.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010674.hg.1 | ENST00000328119.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010674.hg.1 | ENST00000328119.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010674.hg.1 | ENST00000328119.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010674.hg.1 | ENST00000328119.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010674.hg.1 | ENST00000328119.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010674.hg.1 | ENST00000393986.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010674.hg.1 | ENST00000393986.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010674.hg.1 | ENST00000393986.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010674.hg.1 | ENST00000393986.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010674.hg.1 | ENST00000393986.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010674.hg.1 | ENST00000393986.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010674.hg.1 | ENST00000393986.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010674.hg.1 | ENST00000393986.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010674.hg.1 | ENST00000393986.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010674.hg.1 | NM_003771.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010674.hg.1 | NM_003771.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010674.hg.1 | NM_003771.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010674.hg.1 | NM_003771.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010674.hg.1 | NM_003771.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010674.hg.1 | NM_003771.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010674.hg.1 | NM_003771.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010674.hg.1 | NM_003771.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010674.hg.1 | NM_003771.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700184360.hg.1 | ENST00000328119.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184360.hg.1 | ENST00000393986.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184360.hg.1 | NM_003771.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700184360.hg.1 | ENST00000328119.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184360.hg.1 | ENST00000393986.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184360.hg.1 | NM_003771.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700184360.hg.1 | TC1700010674.hg.1 | Clariom D TC |

KRT36
Genomic Map