| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12019503.hg.1 |
ENST00000335999.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12019503.hg.1 |
ENST00000354423.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12019503.hg.1 |
NM_001037806.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12019503.hg.1 |
TC12001482.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1200180976.hg.1 |
ENST00000335999.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200180976.hg.1 |
ENST00000477361.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200180976.hg.1 |
ENST00000491441.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200180976.hg.1 |
NM_001037806.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200180976.hg.1 |
ENST00000335999.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200180976.hg.1 |
ENST00000477361.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200180976.hg.1 |
ENST00000491441.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200180976.hg.1 |
NM_001037806.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200180976.hg.1 |
TC1200010640.hg.1 |
Clariom D TC |