Assay Details
| Gene Symbol: NCKAP5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037806.3 | NP_001032895.2 | 6 - 7 | 555 | 75 | |
| XM_005269050.3 | XP_005269107.1 | 6 - 7 | 880 | 75 | ||
| XM_006719525.2 | XP_006719588.1 | 6 - 7 | 850 | 75 | ||
| XM_011538610.2 | XP_011536912.1 | 7 - 8 | 634 | 75 | ||
| XM_011538611.2 | XP_011536913.1 | 7 - 8 | 604 | 75 | ||
| GenBank mRNA | CN359941.1 | - | 1 - 2 | 68 | 75 | |
| CN359946.1 | - | 6 - 7 | 555 | 75 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001482.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12019497.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12019497.hg.1 | ENST00000354423.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12019497.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12019497.hg.1 | TC12001482.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000433948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000433948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000433948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000433948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000433948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000433948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000433948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000433948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000477361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000477361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000477361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000477361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000477361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000477361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000477361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000477361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000480927.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000480927.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000480927.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000480927.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000491441.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000491441.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000491441.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000491441.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000491441.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000491441.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000491441.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12019495.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12019495.hg.1 | ENST00000354423.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12019495.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12019495.hg.1 | TC12001482.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12019496.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12019496.hg.1 | ENST00000354423.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12019496.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12019496.hg.1 | TC12001482.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200180968.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180968.hg.1 | ENST00000433948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180968.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200180968.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180968.hg.1 | ENST00000433948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180968.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200180968.hg.1 | TC1200010640.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200180969.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180969.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200180969.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180969.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200180969.hg.1 | TC1200010640.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000433948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000433948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000433948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000433948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000433948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000433948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000433948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000433948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000477361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000477361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000477361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000477361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000477361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000477361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000477361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000477361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000480927.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000480927.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000480927.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000480927.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000491441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000491441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000491441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000491441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000491441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000491441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000491441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200180970.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180970.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200180970.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180970.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200180970.hg.1 | TC1200010640.hg.1 | Clariom D TC |

NCKAP5L
Genomic Map