Assay Details
| Gene Symbol: CASC22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135281.1 | 3 | 292 | 82 | ||
| GenBank mRNA | AK093211.1 | - | 3 | 1888 | 82 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102467079 | LOC102467079 | -180.9kb | Anti-sense | Chr. 16 - 52085281 - 52099067 on NCBI Build GRCh38 | |
| Downstream | TOX high mobility group box family member 3 | TOX3 |
CAGF9, TNRC9 |
156.1kb | Sense | Chr. 16 - 52436415 - 52547802 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015671 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015704 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015759 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015673 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555338 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555403 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16896147 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555384 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555393 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015768 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015724 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015758 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555423 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015648 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555288 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015642 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015712 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015730 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015731 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16896153 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555385 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555410 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16896151 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555375 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16896158 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555281 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555412 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015721 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555298 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555408 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555287 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555348 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555295 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555304 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555310 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555344 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015720 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015753 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16896138 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015700 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555394 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015779 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015641 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555277 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015763 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555359 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555414 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015782 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555309 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555343 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555320 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015766 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16896137 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015783 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555279 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16896136 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015647 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555398 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015686 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015789 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015702 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555404 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015675 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555290 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015693 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16896145 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015784 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555368 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555411 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015737 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015715 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555321 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555409 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555357 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015708 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16896156 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015746 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555284 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555302 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015741 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555308 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555413 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555286 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015682 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015707 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015734 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555322 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555416 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015786 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015764 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555400 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16896159 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555377 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555285 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555381 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555395 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555326 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015727 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555336 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16896160 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015785 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555397 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015655 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015718 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555392 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015694 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015729 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015688 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555301 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015676 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555352 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555386 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555396 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555319 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555293 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015698 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015691 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555305 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015662 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555387 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555406 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555327 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015680 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015717 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555315 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015681 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555376 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015695 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015639 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555299 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555306 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555313 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015740 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10015726 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555389 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30555282 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 786785 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 399512 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 598047 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3538104 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 58917 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3538104 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 142265 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P20466235 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03145357 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P702133 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 1556954_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007862.hg.1 | ENST00000569713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007862.hg.1 | ENST00000569713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007862.hg.1 | ENST00000569713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007862.hg.1 | ENST00000628396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007862.hg.1 | ENST00000628396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007862.hg.1 | ENST00000628396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007862.hg.1 | ENST00000628396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007862.hg.1 | ENST00000628396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007862.hg.1 | ENST00000628396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007862.hg.1 | ENST00000569713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007862.hg.1 | ENST00000569713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007862.hg.1 | ENST00000569713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007862.hg.1 | ENST00000628396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007862.hg.1 | ENST00000628396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007862.hg.1 | ENST00000628396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007862.hg.1 | ENST00000628396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007862.hg.1 | ENST00000628396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007862.hg.1 | ENST00000628396.1 | Ensembl |
| Human Genome U133 Plus 2.0 Array | 1556954_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1556954_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556954_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556954_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556954_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556954_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556954_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556954_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556954_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556954_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556954_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556954_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556954_at5 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 139975 | - | - |

CASC22
Genomic Map