Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04005247.hg.1 |
TC04000385.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04005247.hg.1 |
ENST00000399575.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04005247.hg.1 |
ENST00000514338.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04005247.hg.1 |
NM_021225.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400153119.hg.1 |
ENST00000399575.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400153119.hg.1 |
ENST00000505023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400153119.hg.1 |
ENST00000514338.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400153119.hg.1 |
ENST00000615728.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400153119.hg.1 |
NM_001302807.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400153119.hg.1 |
NM_021225.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400153119.hg.1 |
NR_126503.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400153119.hg.1 |
ENST00000399575.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400153119.hg.1 |
ENST00000505023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400153119.hg.1 |
ENST00000514338.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400153119.hg.1 |
ENST00000615728.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400153119.hg.1 |
NM_001302807.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400153119.hg.1 |
NM_021225.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400153119.hg.1 |
NR_126503.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400153119.hg.1 |
TC0400007781.hg.1 |
Clariom D TC |