Assay Details
| Gene Symbol: FSCN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077182.2 | NP_001070650.1 | 1 - 2 | 966 | 71 | |
| NM_012418.3 | NP_036550.1 | 1 - 2 | 966 | 71 | ||
| XM_011524587.2 | XP_011522889.1 | 2 - 3 | 2449 | 71 | ||
| XM_011524590.2 | XP_011522892.1 | 3 - 4 | 4898 | 71 | ||
| GenBank mRNA | AF030165.1 | - | 1 - 2 | 961 | 71 | |
| BC126295.1 | - | 1 - 2 | 882 | 8992011 | 71 | |
| BC130330.1 | - | 1 - 2 | 882 | 40146361 | 71 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000931.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17012246.hg.1 | ENST00000334850.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012246.hg.1 | ENST00000417245.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012246.hg.1 | NM_001077182.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012246.hg.1 | NM_012418.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012246.hg.1 | TC17000931.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009178.hg.1 | ENST00000430912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009178.hg.1 | ENST00000430912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009178.hg.1 | ENST00000442532.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009178.hg.1 | ENST00000442532.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009178.hg.1 | ENST00000442532.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012244.hg.1 | ENST00000417245.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012244.hg.1 | NM_001077182.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012244.hg.1 | NM_012418.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012244.hg.1 | TC17000931.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012247.hg.1 | ENST00000334850.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012247.hg.1 | ENST00000417245.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012247.hg.1 | NM_001077182.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012247.hg.1 | NM_012418.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012247.hg.1 | TC17000931.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009179.hg.1 | ENST00000334850.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009179.hg.1 | ENST00000334850.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009179.hg.1 | ENST00000334850.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009179.hg.1 | ENST00000334850.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009179.hg.1 | ENST00000334850.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009179.hg.1 | ENST00000334850.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009179.hg.1 | ENST00000334850.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009179.hg.1 | ENST00000417245.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009179.hg.1 | ENST00000417245.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009179.hg.1 | ENST00000417245.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009179.hg.1 | ENST00000417245.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009179.hg.1 | ENST00000417245.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009179.hg.1 | ENST00000417245.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009179.hg.1 | ENST00000417245.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009179.hg.1 | ENST00000417245.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009179.hg.1 | ENST00000417245.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009179.hg.1 | ENST00000527221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009179.hg.1 | ENST00000527221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009179.hg.1 | ENST00000527221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009179.hg.1 | ENST00000527221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009179.hg.1 | ENST00000527221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009179.hg.1 | NM_001077182.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009179.hg.1 | NM_001077182.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009179.hg.1 | NM_001077182.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009179.hg.1 | NM_001077182.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009179.hg.1 | NM_001077182.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009179.hg.1 | NM_001077182.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009179.hg.1 | NM_001077182.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009179.hg.1 | NM_001077182.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009179.hg.1 | NM_001077182.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009179.hg.1 | NM_001077182.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009179.hg.1 | NM_012418.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009179.hg.1 | NM_012418.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009179.hg.1 | NM_012418.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009179.hg.1 | NM_012418.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009179.hg.1 | NM_012418.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009179.hg.1 | NM_012418.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009179.hg.1 | NM_012418.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009179.hg.1 | NM_012418.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009179.hg.1 | NM_012418.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012245.hg.1 | ENST00000417245.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012245.hg.1 | NM_001077182.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012245.hg.1 | NM_012418.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012245.hg.1 | TC17000931.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1700169550.hg.1 | ENST00000417245.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169550.hg.1 | NM_001077182.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169550.hg.1 | NM_012418.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169550.hg.1 | ENST00000417245.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169550.hg.1 | NM_001077182.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169550.hg.1 | NM_012418.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169550.hg.1 | TC1700009179.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700169552.hg.1 | ENST00000334850.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169552.hg.1 | ENST00000417245.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169552.hg.1 | NM_001077182.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169552.hg.1 | NM_012418.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169552.hg.1 | ENST00000334850.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169552.hg.1 | ENST00000417245.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169552.hg.1 | NM_001077182.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169552.hg.1 | NM_012418.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169552.hg.1 | TC1700009179.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700169551.hg.1 | ENST00000417245.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169551.hg.1 | NM_001077182.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169551.hg.1 | NM_012418.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169551.hg.1 | ENST00000417245.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169551.hg.1 | NM_001077182.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169551.hg.1 | NM_012418.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169551.hg.1 | TC1700009179.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700009179.hg.1 | ENST00000334850.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009179.hg.1 | ENST00000334850.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009179.hg.1 | ENST00000334850.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009179.hg.1 | ENST00000334850.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009179.hg.1 | ENST00000334850.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009179.hg.1 | ENST00000334850.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009179.hg.1 | ENST00000334850.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009179.hg.1 | ENST00000417245.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009179.hg.1 | ENST00000417245.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009179.hg.1 | ENST00000417245.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009179.hg.1 | ENST00000417245.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009179.hg.1 | ENST00000417245.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009179.hg.1 | ENST00000417245.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009179.hg.1 | ENST00000417245.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009179.hg.1 | ENST00000417245.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009179.hg.1 | ENST00000417245.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009179.hg.1 | ENST00000527221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009179.hg.1 | ENST00000527221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009179.hg.1 | ENST00000527221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009179.hg.1 | ENST00000527221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009179.hg.1 | ENST00000527221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009179.hg.1 | NM_001077182.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009179.hg.1 | NM_001077182.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009179.hg.1 | NM_001077182.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009179.hg.1 | NM_001077182.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009179.hg.1 | NM_001077182.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009179.hg.1 | NM_001077182.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009179.hg.1 | NM_001077182.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009179.hg.1 | NM_001077182.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009179.hg.1 | NM_001077182.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009179.hg.1 | NM_001077182.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009179.hg.1 | NM_012418.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009179.hg.1 | NM_012418.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009179.hg.1 | NM_012418.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009179.hg.1 | NM_012418.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009179.hg.1 | NM_012418.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009179.hg.1 | NM_012418.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009179.hg.1 | NM_012418.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009179.hg.1 | NM_012418.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009179.hg.1 | NM_012418.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009178.hg.1 | ENST00000430912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009178.hg.1 | ENST00000430912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009178.hg.1 | ENST00000442532.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009178.hg.1 | ENST00000442532.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009178.hg.1 | ENST00000442532.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169553.hg.1 | ENST00000334850.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169553.hg.1 | ENST00000417245.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169553.hg.1 | NM_001077182.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169553.hg.1 | NM_012418.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169553.hg.1 | ENST00000334850.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169553.hg.1 | ENST00000417245.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169553.hg.1 | NM_001077182.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169553.hg.1 | NM_012418.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169553.hg.1 | TC1700009179.hg.1 | Clariom D TC |

FSCN2
Genomic Map