Assay Details
| Gene Symbol: FSCN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077182.2 | NP_001070650.1 | 4 - 5 | 1486 | 76 | |
| NM_012418.3 | NP_036550.1 | 4 - 5 | 1414 | 76 | ||
| XM_011524587.2 | XP_011522889.1 | 5 - 6 | 2969 | 76 | ||
| XM_011524590.2 | XP_011522892.1 | 6 - 7 | 5418 | 76 | ||
| GenBank mRNA | AF030165.1 | - | 4 - 5 | 1409 | 76 | |
| AI189621.1 | - | 1 - 2 | 76 | |||
| BC126295.1 | - | 4 - 5 | 1330 | 8992011 | 76 | |
| BC130330.1 | - | 4 - 5 | 1330 | 40146361 | 76 | |
More Information
Set Membership: |
|

FSCN2
Genomic Map