Assay Details
| Gene Symbol: ZSWIM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023072.2 | NP_075560.2 | 5 - 6 | 1205 | 71 | |
| XM_005260023.3 | XP_005260080.1 | 5 - 6 | 1016 | 71 | ||
| XM_017027153.1 | XP_016882642.1 | 5 - 6 | 1239 | 71 | ||
| XM_017027154.1 | XP_016882643.1 | 5 - 6 | 1171 | 71 | ||
| XM_017027156.1 | XP_016882645.1 | 4 - 5 | 743 | 71 | ||
| XM_017027157.1 | XP_016882646.1 | 5 - 6 | 1241 | 71 | ||
| GenBank mRNA | AK024452.1 | - | 5 - 6 | 1205 | 71 | |
| AK299057.1 | - | 3 - 4 | 392 | 71 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000241.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19003553.hg.1 | ENST00000254323.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19003553.hg.1 | ENST00000440752.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19003553.hg.1 | NM_023072.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19003553.hg.1 | TC19000241.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | ENST00000254323.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | ENST00000254323.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | ENST00000254323.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | ENST00000254323.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | ENST00000254323.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | ENST00000254323.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | ENST00000254323.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | ENST00000254323.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | ENST00000254323.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | ENST00000254323.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | ENST00000254323.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | ENST00000254323.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | ENST00000254323.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | ENST00000254323.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | ENST00000254323.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | ENST00000254323.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | ENST00000254323.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | ENST00000254323.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | ENST00000254323.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | ENST00000254323.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | ENST00000590508.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | ENST00000590508.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | ENST00000590508.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | ENST00000590508.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | ENST00000590508.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | ENST00000590508.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | ENST00000590508.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | ENST00000590508.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | ENST00000590508.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | ENST00000590508.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | ENST00000590508.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | ENST00000590508.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | ENST00000590508.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | ENST00000590508.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | ENST00000590508.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | ENST00000590508.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | ENST00000592227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | ENST00000592227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | ENST00000592227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | ENST00000592227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | ENST00000592227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | NM_023072.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | NM_023072.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | NM_023072.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | NM_023072.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | NM_023072.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | NM_023072.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | NM_023072.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | NM_023072.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | NM_023072.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | NM_023072.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | NM_023072.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | NM_023072.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | NM_023072.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | NM_023072.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | NM_023072.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | NM_023072.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | NM_023072.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | NM_023072.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | NM_023072.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007142.hg.1 | NM_023072.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900151460.hg.1 | ENST00000254323.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900151460.hg.1 | ENST00000590508.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900151460.hg.1 | NM_023072.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900151460.hg.1 | TC1900007142.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900151460.hg.1 | ENST00000254323.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900151460.hg.1 | ENST00000590508.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900151460.hg.1 | NM_023072.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | ENST00000254323.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | ENST00000254323.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | ENST00000254323.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | ENST00000254323.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | ENST00000254323.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | ENST00000254323.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | ENST00000254323.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | ENST00000254323.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | ENST00000254323.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | ENST00000254323.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | ENST00000254323.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | ENST00000254323.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | ENST00000254323.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | ENST00000254323.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | ENST00000254323.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | ENST00000254323.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | ENST00000254323.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | ENST00000254323.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | ENST00000254323.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | ENST00000254323.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | ENST00000590508.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | ENST00000590508.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | ENST00000590508.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | ENST00000590508.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | ENST00000590508.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | ENST00000590508.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | ENST00000590508.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | ENST00000590508.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | ENST00000590508.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | ENST00000590508.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | ENST00000590508.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | ENST00000590508.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | ENST00000590508.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | ENST00000590508.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | ENST00000590508.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | ENST00000590508.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | ENST00000592227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | ENST00000592227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | ENST00000592227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | ENST00000592227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | ENST00000592227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | NM_023072.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | NM_023072.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | NM_023072.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | NM_023072.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | NM_023072.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | NM_023072.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | NM_023072.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | NM_023072.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | NM_023072.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | NM_023072.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | NM_023072.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | NM_023072.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | NM_023072.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | NM_023072.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | NM_023072.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | NM_023072.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | NM_023072.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | NM_023072.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | NM_023072.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007142.hg.1 | NM_023072.2 | Refseq |

ZSWIM4
Genomic Map