Assay Details
| Gene Symbol: DCAF15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138353.2 | NP_612362.2 | 11 - 12 | 1635 | 72 | |
| GenBank mRNA | AK093072.1 | - | 11 - 12 | 1652 | 72 | |
| BC002926.2 | - | 9 - 10 | 1389 | 72 | ||
| BC013280.2 | - | 9 - 10 | 1390 | 72 | ||
| BC018894.2 | - | 9 - 10 | 1389 | 72 | ||
| BC018895.2 | - | 9 - 10 | 1389 | 72 | ||
| BC080575.1 | - | 11 - 12 | 1635 | 6580131 | 72 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000248.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000254337.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000254337.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000254337.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000254337.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000254337.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000254337.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000254337.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000254337.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000254337.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000254337.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000254337.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000254337.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000254337.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000254337.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000254337.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000254337.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000254337.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000254337.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000254337.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000585468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000585468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000585468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000585468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000585468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000587307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000587307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000587307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000587307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000587307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000587307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000587307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000587307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000587307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000588523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000588523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000588523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000588523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000588523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000588523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000588523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000588523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000588523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000588523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000588523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000591385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000591385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000591385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000591385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000591802.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000591802.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | ENST00000591802.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | NM_138353.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | NM_138353.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | NM_138353.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | NM_138353.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | NM_138353.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | NM_138353.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | NM_138353.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | NM_138353.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | NM_138353.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | NM_138353.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | NM_138353.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | NM_138353.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | NM_138353.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | NM_138353.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | NM_138353.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | NM_138353.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | NM_138353.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | NM_138353.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007151.hg.1 | NM_138353.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19003626.hg.1 | ENST00000254337.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19003626.hg.1 | NM_138353.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19003626.hg.1 | TC19000248.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000254337.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000254337.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000254337.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000254337.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000254337.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000254337.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000254337.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000254337.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000254337.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000254337.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000254337.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000254337.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000254337.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000254337.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000254337.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000254337.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000254337.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000254337.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000254337.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000585468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000585468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000585468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000585468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000585468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000587307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000587307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000587307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000587307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000587307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000587307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000587307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000587307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000587307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000588523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000588523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000588523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000588523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000588523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000588523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000588523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000588523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000588523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000588523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000588523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000591385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000591385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000591385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000591385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000591802.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000591802.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | ENST00000591802.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | NM_138353.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | NM_138353.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | NM_138353.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | NM_138353.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | NM_138353.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | NM_138353.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | NM_138353.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | NM_138353.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | NM_138353.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | NM_138353.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | NM_138353.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | NM_138353.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | NM_138353.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | NM_138353.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | NM_138353.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | NM_138353.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | NM_138353.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | NM_138353.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007151.hg.1 | NM_138353.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900151589.hg.1 | ENST00000254337.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900151589.hg.1 | ENST00000587307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900151589.hg.1 | ENST00000588523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900151589.hg.1 | NM_138353.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900151589.hg.1 | TC1900007151.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900151589.hg.1 | ENST00000254337.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900151589.hg.1 | ENST00000587307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900151589.hg.1 | ENST00000588523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900151589.hg.1 | NM_138353.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900151588.hg.1 | ENST00000254337.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900151588.hg.1 | ENST00000587307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900151588.hg.1 | ENST00000588523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900151588.hg.1 | ENST00000591385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900151588.hg.1 | NM_138353.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900151588.hg.1 | TC1900007151.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900151588.hg.1 | ENST00000254337.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900151588.hg.1 | ENST00000587307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900151588.hg.1 | ENST00000588523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900151588.hg.1 | ENST00000591385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900151588.hg.1 | NM_138353.3 | Refseq |

DCAF15
Genomic Map