Assay Details
| Gene Symbol: ITGB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000212.2 | NP_000203.2 | 11 - 12 | 1929 | 66 | |
| GenBank mRNA | BC127666.1 | - | 11 - 12 | 1929 | 40128462 | 66 |
| BC127667.1 | - | 11 - 12 | 1933 | 40128464 | 66 | |
| KC120777.1 | - | 11 - 12 | 1930 | 66 | ||
| KC120778.1 | - | 11 - 12 | 1929 | 66 | ||
| M35999.1 | - | 11 - 12 | 1927 | 66 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17002878.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of protein phosphorylation
positive regulation of endothelial cell proliferation
platelet degranulation
cell-substrate junction assembly
cell adhesion
cell-matrix adhesion
integrin-mediated signaling pathway
blood coagulation
positive regulation of endothelial cell migration
negative regulation of macrophage derived foam cell differentiation
negative regulation of lipid storage
smooth muscle cell migration
cell growth
cell migration
platelet activation
extracellular matrix organization
positive regulation of vascular endothelial growth factor receptor signaling pathway
cell-substrate adhesion
activation of protein kinase activity
negative regulation of lipid transport
heterotypic cell-cell adhesion
substrate adhesion-dependent cell spreading
tube development
apolipoprotein A-I-mediated signaling pathway
wound healing
regulation of bone resorption
negative regulation of low-density lipoprotein particle receptor biosynthetic process
viral entry into host cell
vascular endothelial growth factor receptor signaling pathway
mesodermal cell differentiation
positive regulation of peptidyl-tyrosine phosphorylation
negative regulation of lipoprotein metabolic process
leukocyte migration
negative chemotaxis
angiogenesis involved in wound healing
platelet aggregation
virus receptor activity
fibronectin binding
protease binding
protein disulfide isomerase activity
platelet-derived growth factor receptor binding
protein binding
coreceptor activity
fibroblast growth factor binding
enzyme binding
C-X3-C chemokine binding
insulin-like growth factor I binding
neuregulin binding
identical protein binding
vascular endothelial growth factor receptor 2 binding
cell adhesion molecule binding
extracellular matrix binding
positive regulation of protein phosphorylation
positive regulation of endothelial cell proliferation
platelet degranulation
cell-substrate junction assembly
cell adhesion
cell-matrix adhesion
integrin-mediated signaling pathway
blood coagulation
positive regulation of endothelial cell migration
negative regulation of macrophage derived foam cell differentiation
negative regulation of lipid storage
smooth muscle cell migration
cell growth
cell migration
platelet activation
extracellular matrix organization
positive regulation of vascular endothelial growth factor receptor signaling pathway
cell-substrate adhesion
activation of protein kinase activity
negative regulation of lipid transport
heterotypic cell-cell adhesion
substrate adhesion-dependent cell spreading
tube development
apolipoprotein A-I-mediated signaling pathway
wound healing
regulation of bone resorption
negative regulation of low-density lipoprotein particle receptor biosynthetic process
viral entry into host cell
vascular endothelial growth factor receptor signaling pathway
mesodermal cell differentiation
positive regulation of peptidyl-tyrosine phosphorylation
negative regulation of lipoprotein metabolic process
leukocyte migration
negative chemotaxis
angiogenesis involved in wound healing
platelet aggregation
virus receptor activity
fibronectin binding
protease binding
protein disulfide isomerase activity
platelet-derived growth factor receptor binding
protein binding
coreceptor activity
fibroblast growth factor binding
enzyme binding
C-X3-C chemokine binding
insulin-like growth factor I binding
neuregulin binding
identical protein binding
vascular endothelial growth factor receptor 2 binding
cell adhesion molecule binding
extracellular matrix binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17007428.hg.1 | TC17002878.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007428.hg.1 | ENST00000262017.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007428.hg.1 | ENST00000559488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007428.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | ENST00000559488.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | ENST00000559488.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | ENST00000559488.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | ENST00000559488.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | ENST00000559488.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | ENST00000559488.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | ENST00000559488.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | ENST00000559488.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | ENST00000559488.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | ENST00000559488.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | ENST00000559488.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | ENST00000559488.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | ENST00000559488.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | ENST00000559488.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | ENST00000559488.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | ENST00000559488.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | ENST00000559488.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | ENST00000559488.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | ENST00000559488.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | ENST00000571680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | ENST00000571680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | ENST00000571680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | ENST00000571680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | ENST00000571680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | ENST00000571680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | ENST00000571680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | ENST00000571680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | ENST00000571680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | ENST00000571680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | ENST00000571680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | ENST00000571680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | ENST00000571680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | ENST00000573377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | ENST00000573377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | ENST00000573377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | ENST00000573377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012274.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007429.hg.1 | TC17002878.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007429.hg.1 | ENST00000262017.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007429.hg.1 | ENST00000559488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007429.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700202763.hg.1 | ENST00000559488.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700202763.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700202763.hg.1 | ENST00000559488.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700202763.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700202763.hg.1 | TC1700012274.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700202764.hg.1 | ENST00000559488.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700202764.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700202764.hg.1 | ENST00000559488.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700202764.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700202764.hg.1 | TC1700012274.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | ENST00000559488.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | ENST00000559488.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | ENST00000559488.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | ENST00000559488.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | ENST00000559488.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | ENST00000559488.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | ENST00000559488.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | ENST00000559488.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | ENST00000559488.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | ENST00000559488.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | ENST00000559488.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | ENST00000559488.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | ENST00000559488.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | ENST00000559488.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | ENST00000559488.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | ENST00000559488.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | ENST00000559488.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | ENST00000559488.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | ENST00000559488.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | ENST00000571680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | ENST00000571680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | ENST00000571680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | ENST00000571680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | ENST00000571680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | ENST00000571680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | ENST00000571680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | ENST00000571680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | ENST00000571680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | ENST00000571680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | ENST00000571680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | ENST00000571680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | ENST00000571680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | ENST00000573377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | ENST00000573377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | ENST00000573377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | ENST00000573377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | NM_000212.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012274.hg.1 | NM_000212.2 | Refseq |

ITGB3
Genomic Map