Assay Details
| Gene Symbol: KANSL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005263982.2 | XP_005264039.1 | 21 - 22 | 2844 | 125 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KANSL3| Gene Symbol: KANSL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005263982.2 | XP_005264039.1 | 21 - 22 | 2844 | 125 | |
Set Membership: |
|