Assay Details
| Gene Symbol: KBTBD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318724.1 | NP_001305653.1 | 1 - 2 | 192 | 59 | |
| GenBank mRNA | DA170281.1 | - | 1 - 2 | 147 | 59 | |
| DA883418.1 | - | 1 - 2 | 129 | 59 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the NM_001318724.1 transcripts.
This assay is in the 5'UTR region of the NM_001318724.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000395288.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000395288.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000395288.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000395288.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000395288.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000395288.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000395288.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000395288.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000395288.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000395288.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000430070.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000430070.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000430070.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000430070.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000430070.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000430070.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000430070.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000430070.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000430070.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000525720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000525720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000525720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000525720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000525720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000525720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000525720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000525720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000525720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000526005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000526005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000526005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000526005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000526005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000526005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000526005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000526005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000526005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000526005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000529499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000529499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000529499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000529499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000529499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000529499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000529946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000529946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000529946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000530668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000530668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000530668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000530668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000530668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000530668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000531067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000531067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000531067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000531067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000533290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000533290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000533290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000533290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000533290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000533290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000533290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000533290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000533290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000533290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000533290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000533290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000533290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000534239.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000534239.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | ENST00000534239.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | NM_016506.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | NM_016506.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | NM_016506.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | NM_016506.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | NM_016506.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | NM_016506.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | NM_016506.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | NM_016506.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | NM_016506.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | NM_016506.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | NM_018095.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | NM_018095.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | NM_018095.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | NM_018095.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | NM_018095.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | NM_018095.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | NM_018095.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | NM_018095.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | NM_018095.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010751.hg.1 | NM_018095.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000395288.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000395288.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000395288.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000395288.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000395288.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000395288.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000395288.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000395288.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000395288.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000395288.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000430070.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000430070.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000430070.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000430070.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000430070.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000430070.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000430070.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000430070.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000430070.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000525720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000525720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000525720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000525720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000525720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000525720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000525720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000525720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000525720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000526005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000526005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000526005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000526005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000526005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000526005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000526005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000526005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000526005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000526005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000529499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000529499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000529499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000529499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000529499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000529499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000529946.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000529946.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000529946.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000530668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000530668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000530668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000530668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000530668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000530668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000531067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000531067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000531067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000531067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000533290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000533290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000533290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000533290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000533290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000533290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000533290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000533290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000533290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000533290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000533290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000533290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000533290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000534239.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000534239.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | ENST00000534239.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | NM_016506.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | NM_016506.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | NM_016506.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | NM_016506.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | NM_016506.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | NM_016506.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | NM_016506.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | NM_016506.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | NM_016506.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | NM_016506.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | NM_018095.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | NM_018095.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | NM_018095.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | NM_018095.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | NM_018095.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | NM_018095.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | NM_018095.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | NM_018095.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | NM_018095.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010751.hg.1 | NM_018095.5 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 280200 | - | - |

KBTBD4
Genomic Map