Assay Details
| Gene Symbol: TH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199292.2 | NP_954986.2 | 1 - 2 | 121 | 137 | |
| GenBank mRNA | M17589.1 | - | 1 - 2 | 111 | 137 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: TH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199292.2 | NP_954986.2 | 1 - 2 | 121 | 137 | |
| GenBank mRNA | M17589.1 | - | 1 - 2 | 111 | 137 | |
Set Membership: |
|