Assay Details
| Gene Symbol: TH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011520335.2 | XP_011518637.1 | 1 - 2 | 2393 | 53 | |
| GenBank mRNA | BC143614.1 | - | 1 - 2 | 189 | 9052127 | 53 |
| DQ677337.1 | - | 1 - 2 | 107 | 53 | ||
More Information
Set Membership: |
|

TH
Genomic Map