Assay Details
| Gene Symbol: CALY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001321617.1 | NP_001308546.1 | 4 - 5 | 533 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001321617.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000368555.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000368555.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000368555.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000368555.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000467433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000467433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000467433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000467433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000467433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000467433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000467611.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000467611.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000467611.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000467611.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000467611.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000467611.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000467611.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000605518.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000368555.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000368555.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000368555.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000368555.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000467433.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000467433.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000467433.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000467433.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000467433.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000467433.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000467611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000467611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000467611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000467611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000467611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000467611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000467611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000605518.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | NM_015722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | NM_015722.3 | Refseq |

CALY
Genomic Map