Assay Details
| Gene Symbol: CDC37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007065.3 | NP_008996.1 | 2 - 3 | 494 | 61 | |
| GenBank mRNA | BC000083.2 | - | 2 - 3 | 418 | 3505011 | 61 |
| BC008793.2 | - | 2 - 3 | 418 | 3619769 | 61 | |
| BE796008.1 | - | 2 - 3 | 498 | 61 | ||
| BT006796.1 | - | 2 - 3 | 378 | 61 | ||
| CR542249.1 | - | 2 - 3 | 378 | 61 | ||
| U43077.1 | - | 2 - 3 | 386 | 61 | ||
| U63131.1 | - | 2 - 3 | 426 | 61 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001165.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cyclin-dependent protein serine/threonine kinase activity
protein folding
protein targeting
posttranscriptional regulation of gene expression
peptidyl-tyrosine phosphorylation
ERBB2 signaling pathway
protein stabilization
regulation of interferon-gamma-mediated signaling pathway
regulation of type I interferon-mediated signaling pathway
mitophagy in response to mitochondrial depolarization
protein tyrosine kinase activity
protein binding
protein kinase regulator activity
kinase binding
protein kinase binding
heat shock protein binding
unfolded protein binding
chaperone binding
Hsp90 protein binding
regulation of cyclin-dependent protein serine/threonine kinase activity
protein folding
protein targeting
posttranscriptional regulation of gene expression
peptidyl-tyrosine phosphorylation
ERBB2 signaling pathway
protein stabilization
regulation of interferon-gamma-mediated signaling pathway
regulation of type I interferon-mediated signaling pathway
mitophagy in response to mitochondrial depolarization
protein tyrosine kinase activity
protein binding
protein kinase regulator activity
kinase binding
protein kinase binding
heat shock protein binding
unfolded protein binding
chaperone binding
Hsp90 protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500012620.hg.1 | ENST00000519230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000222005.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000222005.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000222005.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000222005.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000222005.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000222005.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000222005.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000222005.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000222005.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000222005.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000222005.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000222005.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000222005.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000222005.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000222005.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000222005.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000408639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000588498.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000588498.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000588498.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000588498.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000588498.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000588498.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000588498.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000588498.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000588498.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000588869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000588869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000588869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000588869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000588869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000588869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000588869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000588869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000588869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000588869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000588869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000589331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000589331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000589331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000589625.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000589625.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000589625.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000589625.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000589625.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000589625.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000589625.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000589629.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000589629.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000589629.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000589629.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000589629.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000589629.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000589629.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000589629.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000589755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000589755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000589755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000589755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000589755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000589755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000589755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000589755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000589755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000590632.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000590632.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000590632.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000590632.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000591248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000591248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000591248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000591248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000591248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000591248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000591248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000591248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000591248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000591248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000591248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000591248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000591248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000591248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000591248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000591248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000593124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000593124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000593124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000593124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000593124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000593124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000593124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000593124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000593124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000593124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000593124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000593124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000593124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000593124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | ENST00000593124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | NM_007065.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | NM_007065.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | NM_007065.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | NM_007065.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | NM_007065.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | NM_007065.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | NM_007065.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | NM_007065.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | NM_007065.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | NM_007065.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | NM_007065.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | NM_007065.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | NM_007065.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | NM_007065.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | NM_007065.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | NM_007065.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | NM_007065.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009628.hg.1 | NR_031592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19016232.hg.1 | ENST00000222005.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19016232.hg.1 | ENST00000408639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19016232.hg.1 | NM_007065.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19016232.hg.1 | NR_031592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19016232.hg.1 | TC19001165.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0500012620.hg.1 | ENST00000519230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900177954.hg.1 | ENST00000222005.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900177954.hg.1 | ENST00000408639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900177954.hg.1 | ENST00000588869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900177954.hg.1 | ENST00000589629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900177954.hg.1 | ENST00000591248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900177954.hg.1 | ENST00000593124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900177954.hg.1 | NM_007065.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900177954.hg.1 | NR_031592.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900177954.hg.1 | TC1900009628.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900177954.hg.1 | ENST00000222005.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900177954.hg.1 | ENST00000408639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900177954.hg.1 | ENST00000588869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900177954.hg.1 | ENST00000589629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900177954.hg.1 | ENST00000591248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900177954.hg.1 | ENST00000593124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900177954.hg.1 | NM_007065.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900177954.hg.1 | NR_031592.1 | Refseq |

CDC37
Genomic Map