Assay Details
| Gene Symbol: CDK5RAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011528860.1 | XP_011527162.1 | 3 - 4 | 230 | 74 | |
| XM_017027876.1 | XP_016883365.1 | 5 - 6 | 669 | 74 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011528860.1, XM_017027876.1 transcripts.
Set Membership: |
|

CDK5RAP1
Genomic Map