Assay Details
| Gene Symbol: ATP6V1G3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133326.1 | NP_579872.1 | 3 - 4 | 231 | 88 | |
| XM_011509187.2 | XP_011507489.1 | 2 - 3 | 256 | 88 | ||
| GenBank mRNA | BC101131.2 | - | 2 - 3 | 151 | 40014383 | 88 |
| BF509031.1 | - | 2 - 3 | 88 | |||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003667.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100016851.hg.1 | ENST00000281087.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016851.hg.1 | ENST00000281087.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016851.hg.1 | ENST00000281087.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016851.hg.1 | ENST00000281087.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016851.hg.1 | ENST00000281087.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016851.hg.1 | ENST00000281087.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016851.hg.1 | ENST00000281087.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016851.hg.1 | ENST00000309309.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016851.hg.1 | ENST00000309309.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016851.hg.1 | ENST00000309309.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016851.hg.1 | ENST00000309309.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016851.hg.1 | ENST00000309309.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016851.hg.1 | ENST00000309309.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016851.hg.1 | ENST00000309309.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016851.hg.1 | ENST00000309309.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016851.hg.1 | ENST00000367382.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016851.hg.1 | ENST00000367382.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016851.hg.1 | ENST00000367382.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016851.hg.1 | ENST00000367382.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016851.hg.1 | ENST00000367382.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016851.hg.1 | ENST00000367382.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016851.hg.1 | ENST00000367382.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016851.hg.1 | ENST00000489986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016851.hg.1 | ENST00000489986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016851.hg.1 | ENST00000489986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016851.hg.1 | ENST00000489986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016851.hg.1 | ENST00000489986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016851.hg.1 | NM_133262.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016851.hg.1 | NM_133262.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016851.hg.1 | NM_133262.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016851.hg.1 | NM_133262.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016851.hg.1 | NM_133262.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016851.hg.1 | NM_133262.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016851.hg.1 | NM_133262.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016851.hg.1 | NM_133326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016851.hg.1 | NM_133326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016851.hg.1 | NM_133326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016851.hg.1 | NM_133326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016851.hg.1 | NM_133326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016851.hg.1 | NM_133326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016851.hg.1 | NM_133326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016851.hg.1 | NM_133326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056263.hg.1 | TC01003667.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056263.hg.1 | ENST00000281087.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056263.hg.1 | ENST00000309309.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056263.hg.1 | ENST00000367381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056263.hg.1 | ENST00000367382.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056263.hg.1 | ENST00000489986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056263.hg.1 | NM_133262.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056263.hg.1 | NM_133326.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100227523.hg.1 | ENST00000281087.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100227523.hg.1 | ENST00000309309.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100227523.hg.1 | ENST00000367382.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100227523.hg.1 | ENST00000489986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100227523.hg.1 | NM_133262.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100227523.hg.1 | NM_133326.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100227523.hg.1 | ENST00000281087.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100227523.hg.1 | ENST00000309309.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100227523.hg.1 | ENST00000367382.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100227523.hg.1 | ENST00000489986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100227523.hg.1 | NM_133262.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100227523.hg.1 | NM_133326.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100227523.hg.1 | TC0100016851.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100227524.hg.1 | ENST00000281087.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100227524.hg.1 | ENST00000309309.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100227524.hg.1 | ENST00000367382.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100227524.hg.1 | ENST00000489986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100227524.hg.1 | NM_133262.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100227524.hg.1 | NM_133326.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100227524.hg.1 | ENST00000281087.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100227524.hg.1 | ENST00000309309.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100227524.hg.1 | ENST00000367382.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100227524.hg.1 | ENST00000489986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100227524.hg.1 | NM_133262.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100227524.hg.1 | NM_133326.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100227524.hg.1 | TC0100016851.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100016851.hg.1 | ENST00000281087.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016851.hg.1 | ENST00000281087.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016851.hg.1 | ENST00000281087.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016851.hg.1 | ENST00000281087.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016851.hg.1 | ENST00000281087.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016851.hg.1 | ENST00000281087.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016851.hg.1 | ENST00000281087.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016851.hg.1 | ENST00000309309.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016851.hg.1 | ENST00000309309.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016851.hg.1 | ENST00000309309.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016851.hg.1 | ENST00000309309.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016851.hg.1 | ENST00000309309.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016851.hg.1 | ENST00000309309.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016851.hg.1 | ENST00000309309.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016851.hg.1 | ENST00000309309.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016851.hg.1 | ENST00000367382.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016851.hg.1 | ENST00000367382.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016851.hg.1 | ENST00000367382.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016851.hg.1 | ENST00000367382.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016851.hg.1 | ENST00000367382.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016851.hg.1 | ENST00000367382.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016851.hg.1 | ENST00000367382.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016851.hg.1 | ENST00000489986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016851.hg.1 | ENST00000489986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016851.hg.1 | ENST00000489986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016851.hg.1 | ENST00000489986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016851.hg.1 | ENST00000489986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016851.hg.1 | NM_133262.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016851.hg.1 | NM_133262.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016851.hg.1 | NM_133262.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016851.hg.1 | NM_133262.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016851.hg.1 | NM_133262.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016851.hg.1 | NM_133262.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016851.hg.1 | NM_133262.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016851.hg.1 | NM_133326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016851.hg.1 | NM_133326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016851.hg.1 | NM_133326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016851.hg.1 | NM_133326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016851.hg.1 | NM_133326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016851.hg.1 | NM_133326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016851.hg.1 | NM_133326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016851.hg.1 | NM_133326.1 | Refseq |

ATP6V1G3
Genomic Map