Assay Details
| Gene Symbol: PARP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042618.1 | NP_001036083.1 | 2 - 3 | 227 | 86 | |
| XM_017020912.1 | XP_016876401.1 | 2 - 3 | 800 | 86 | ||
| GenBank mRNA | AJ236912.1 | - | 2 - 3 | 201 | 86 | |
| AK290079.1 | - | 2 - 3 | 206 | 86 | ||
| AK300971.1 | - | 2 - 3 | 206 | 86 | ||
More Information
Set Membership: |
|

PARP2
Genomic Map